Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35563209-35563466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM166B
Entrez Gene ID
730112
Gene Name
family with sequence similarity 166 member B
Gene Aliases
-
UniGene
Hs.98959
Gene Chromosome Location
Chr.9: 35561830-35563899 on Build GRCh38
Transcripts
NM_001099951.3 NM_001164310.2 NM_001287238.1 NM_001287239.1 XM_011518028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927459 c.185C>T p.P62L Chr.9: 35563267-35563267 on Build GRCh38
4946607 c.242G>T p.R81L Chr.9: 35563210-35563210 on Build GRCh38
2774268 c.69T>C p.T23T Chr.9: 35563383-35563383 on Build GRCh38
1151270 c.222G>T p.R74S Chr.9: 35563230-35563230 on Build GRCh38
6332069 c.174C>T p.R58R Chr.9: 35563278-35563278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772678302 Chr.9: 35563329-35563349 on Build GRCh38
rs761750554 Chr.9: 35563421-35563421 on Build GRCh38
rs746933959 Chr.9: 35563399-35563399 on Build GRCh38
rs925187272 Chr.9: 35563425-35563425 on Build GRCh38
rs367759645 Chr.9: 35563365-35563365 on Build GRCh38
rs374415795 Chr.9: 35563315-35563315 on Build GRCh38
rs571998273 Chr.9: 35563283-35563283 on Build GRCh38
rs374415250 Chr.9: 35563355-35563355 on Build GRCh38
rs936595904 Chr.9: 35563432-35563432 on Build GRCh38
rs775347237 Chr.9: 35563342-35563342 on Build GRCh38
rs780659260 Chr.9: 35563279-35563279 on Build GRCh38
rs767478334 Chr.9: 35563233-35563233 on Build GRCh38
rs745533992 Chr.9: 35563383-35563383 on Build GRCh38
rs966511399 Chr.9: 35563377-35563377 on Build GRCh38
rs774162238 Chr.9: 35563328-35563328 on Build GRCh38
rs750680581 Chr.9: 35563358-35563358 on Build GRCh38
rs756819931 Chr.9: 35563378-35563378 on Build GRCh38
rs894534175 Chr.9: 35563277-35563277 on Build GRCh38
rs759282340 Chr.9: 35563332-35563332 on Build GRCh38
rs754023113 Chr.9: 35563255-35563255 on Build GRCh38
rs779315158 Chr.9: 35563259-35563259 on Build GRCh38
rs1004823684 Chr.9: 35563287-35563287 on Build GRCh38
rs771876291 Chr.9: 35563337-35563337 on Build GRCh38
rs760608891 Chr.9: 35563344-35563344 on Build GRCh38
rs758865797 Chr.9: 35563264-35563264 on Build GRCh38
rs762342046 Chr.9: 35563287-35563288 on Build GRCh38
rs370308626 Chr.9: 35563382-35563382 on Build GRCh38
rs775330386 Chr.9: 35563392-35563392 on Build GRCh38
rs201290396 Chr.9: 35563363-35563363 on Build GRCh38
rs770536033 Chr.9: 35563321-35563321 on Build GRCh38
rs771618937 Chr.9: 35563316-35563316 on Build GRCh38
rs370675504 Chr.9: 35563280-35563280 on Build GRCh38
rs755602578 Chr.9: 35563367-35563367 on Build GRCh38
rs764171830 Chr.9: 35563250-35563250 on Build GRCh38
rs201489188 Chr.9: 35563316-35563316 on Build GRCh38
rs187370786 Chr.9: 35563248-35563248 on Build GRCh38
rs765372004 Chr.9: 35563427-35563427 on Build GRCh38
rs377446974 Chr.9: 35563356-35563356 on Build GRCh38
rs748786080 Chr.9: 35563370-35563370 on Build GRCh38
rs75679360 Chr.9: 35563364-35563364 on Build GRCh38
rs777217905 Chr.9: 35563298-35563298 on Build GRCh38
rs759040201 Chr.9: 35563291-35563300 on Build GRCh38
rs768518423 Chr.9: 35563402-35563402 on Build GRCh38
rs760385725 Chr.9: 35563396-35563397 on Build GRCh38
rs771788149 Chr.9: 35563386-35563386 on Build GRCh38
rs774899761 Chr.9: 35563402-35563403 on Build GRCh38
rs774726113 Chr.9: 35563271-35563283 on Build GRCh38
rs553046884 Chr.9: 35563405-35563405 on Build GRCh38
rs1015845552 Chr.9: 35563297-35563297 on Build GRCh38
rs907356284 Chr.9: 35563286-35563286 on Build GRCh38
rs1054869980 Chr.9: 35563262-35563262 on Build GRCh38
rs376506269 Chr.9: 35563258-35563258 on Build GRCh38
rs752658080 Chr.9: 35563238-35563238 on Build GRCh38
rs750795243 Chr.9: 35563260-35563260 on Build GRCh38
rs764228168 Chr.9: 35563350-35563350 on Build GRCh38
rs572833588 Chr.9: 35563431-35563431 on Build GRCh38

View Full Product Details