Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 63147216-63147489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1A
Entrez Gene ID
552
Gene Name
arginine vasopressin receptor 1A
Gene Aliases
AVPR V1a, AVPR1, V1aR
UniGene
Hs.2131
Gene Chromosome Location
Chr.12: 63142759-63152810 on Build GRCh38
Transcripts
NM_000706.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5462254 c.1216A>G p.K406E Chr.12: 63147400-63147400 on Build GRCh38
3464152 c.1207G>A p.D403N Chr.12: 63147409-63147409 on Build GRCh38
3464151 c.1246G>A p.V416I Chr.12: 63147370-63147370 on Build GRCh38
3872237 c.1151G>A p.R384K Chr.12: 63147465-63147465 on Build GRCh38
137584 c.1174C>T p.R392* Chr.12: 63147442-63147442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886196587 Chr.12: 63147449-63147449 on Build GRCh38
rs1037957085 Chr.12: 63147456-63147456 on Build GRCh38
rs748889364 Chr.12: 63147316-63147316 on Build GRCh38
rs771813628 Chr.12: 63147413-63147413 on Build GRCh38
rs755580921 Chr.12: 63147371-63147371 on Build GRCh38
rs144329263 Chr.12: 63147377-63147377 on Build GRCh38
rs780873156 Chr.12: 63147357-63147357 on Build GRCh38
rs750951531 Chr.12: 63147444-63147444 on Build GRCh38
rs61734518 Chr.12: 63147420-63147420 on Build GRCh38
rs761842296 Chr.12: 63147442-63147442 on Build GRCh38
rs558973145 Chr.12: 63147302-63147302 on Build GRCh38
rs35856440 Chr.12: 63147283-63147284 on Build GRCh38
rs770889859 Chr.12: 63147393-63147393 on Build GRCh38
rs941750571 Chr.12: 63147450-63147451 on Build GRCh38
rs374551459 Chr.12: 63147452-63147452 on Build GRCh38
rs373731451 Chr.12: 63147343-63147343 on Build GRCh38
rs865851696 Chr.12: 63147421-63147421 on Build GRCh38
rs758727650 Chr.12: 63147353-63147353 on Build GRCh38
rs778784149 Chr.12: 63147402-63147402 on Build GRCh38
rs143654785 Chr.12: 63147423-63147423 on Build GRCh38
rs762419353 Chr.12: 63147405-63147405 on Build GRCh38
rs757791989 Chr.12: 63147323-63147323 on Build GRCh38
rs192411187 Chr.12: 63147345-63147345 on Build GRCh38
rs775808648 Chr.12: 63147422-63147422 on Build GRCh38
rs923449778 Chr.12: 63147259-63147259 on Build GRCh38
rs745647513 Chr.12: 63147404-63147404 on Build GRCh38
rs776834928 Chr.12: 63147440-63147440 on Build GRCh38
rs749166815 Chr.12: 63147392-63147392 on Build GRCh38
rs910225635 Chr.12: 63147354-63147354 on Build GRCh38
rs752367784 Chr.12: 63147363-63147363 on Build GRCh38
rs774852140 Chr.12: 63147314-63147314 on Build GRCh38
rs527913816 Chr.12: 63147300-63147300 on Build GRCh38

View Full Product Details