Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 89694960-89695213 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK10
Entrez Gene ID
8558
Gene Name
cyclin dependent kinase 10
Gene Aliases
PISSLRE
UniGene
Hs.699177
Gene Chromosome Location
Chr.16: 89686612-89696364 on Build GRCh38
Transcripts
NM_001098533.2 NM_001160367.1 NM_052987.3 NM_052988.4 NR_027702.1 NR_027703.1 XM_006721308.2 XM_006721310.2 XM_011523405.2 XM_011523406.2 XM_011523407.2 XM_011523408.2 XM_011523410.2 XM_011523411.2 XM_011523412.1 XM_011523413.2 XM_011523414.2 XM_011523415.2 XM_011523416.2 XM_017023806.1 XM_017023807.1 XM_017023808.1 XM_017023809.1 XM_017023810.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4063634 c.874C>G p.L292V Chr.16: 89695012-89695012 on Build GRCh38
6002409 c.665C>T p.S222L Chr.16: 89695016-89695016 on Build GRCh38
7360613 c.932G>A p.R311K Chr.16: 89695070-89695070 on Build GRCh38
4141955 c.861C>T p.H287H Chr.16: 89694999-89694999 on Build GRCh38
6351755 c.822G>A p.Q274Q Chr.16: 89694960-89694960 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775925838 Chr.16: 89695016-89695016 on Build GRCh38
rs890508691 Chr.16: 89695084-89695084 on Build GRCh38
rs1000629665 Chr.16: 89695108-89695108 on Build GRCh38
rs2235129 Chr.16: 89695183-89695183 on Build GRCh38
rs764716288 Chr.16: 89695023-89695023 on Build GRCh38
rs746006476 Chr.16: 89695059-89695059 on Build GRCh38
rs777674394 Chr.16: 89695099-89695099 on Build GRCh38
rs555214500 Chr.16: 89695087-89695087 on Build GRCh38
rs373606849 Chr.16: 89695080-89695080 on Build GRCh38
rs775664961 Chr.16: 89695064-89695064 on Build GRCh38
rs546017098 Chr.16: 89695171-89695171 on Build GRCh38
rs773229157 Chr.16: 89694987-89694987 on Build GRCh38
rs766960979 Chr.16: 89695008-89695009 on Build GRCh38
rs370403313 Chr.16: 89695053-89695053 on Build GRCh38
rs368047070 Chr.16: 89695086-89695086 on Build GRCh38
rs951388744 Chr.16: 89695079-89695079 on Build GRCh38
rs531781901 Chr.16: 89695086-89695087 on Build GRCh38
rs1024766354 Chr.16: 89695169-89695169 on Build GRCh38
rs945741259 Chr.16: 89695144-89695144 on Build GRCh38
rs572011989 Chr.16: 89695150-89695150 on Build GRCh38
rs760306947 Chr.16: 89695077-89695078 on Build GRCh38
rs942868530 Chr.16: 89694987-89694988 on Build GRCh38
rs1045488108 Chr.16: 89695040-89695040 on Build GRCh38
rs774549236 Chr.16: 89695057-89695057 on Build GRCh38
rs983176488 Chr.16: 89695174-89695174 on Build GRCh38
rs367782553 Chr.16: 89695089-89695089 on Build GRCh38
rs552018497 Chr.16: 89695098-89695098 on Build GRCh38
rs199534729 Chr.16: 89695103-89695103 on Build GRCh38
rs900773343 Chr.16: 89695032-89695032 on Build GRCh38
rs761144743 Chr.16: 89695068-89695068 on Build GRCh38
rs771111623 Chr.16: 89694999-89694999 on Build GRCh38
rs761026492 Chr.16: 89695021-89695021 on Build GRCh38
rs879562633 Chr.16: 89695060-89695060 on Build GRCh38
rs767833852 Chr.16: 89695010-89695010 on Build GRCh38
rs200702119 Chr.16: 89695067-89695067 on Build GRCh38
rs755964645 Chr.16: 89695050-89695050 on Build GRCh38
rs932133225 Chr.16: 89695056-89695056 on Build GRCh38
rs907552522 Chr.16: 89695024-89695024 on Build GRCh38
rs376865715 Chr.16: 89695031-89695031 on Build GRCh38
rs752176667 Chr.16: 89695044-89695044 on Build GRCh38
rs769784266 Chr.16: 89694982-89694982 on Build GRCh38
rs1039975251 Chr.16: 89694996-89694996 on Build GRCh38
rs936960046 Chr.16: 89695125-89695125 on Build GRCh38
rs749925375 Chr.16: 89695026-89695026 on Build GRCh38
rs942246249 Chr.16: 89695030-89695030 on Build GRCh38
rs1032258071 Chr.16: 89695124-89695124 on Build GRCh38
rs564485825 Chr.16: 89695182-89695182 on Build GRCh38
rs756687313 Chr.16: 89695165-89695165 on Build GRCh38
rs772349612 Chr.16: 89695061-89695061 on Build GRCh38
rs534464908 Chr.16: 89695115-89695115 on Build GRCh38
rs912885975 Chr.16: 89695135-89695135 on Build GRCh38
rs539203490 Chr.16: 89695027-89695027 on Build GRCh38
rs552795799 Chr.16: 89695129-89695129 on Build GRCh38
rs747970770 Chr.16: 89694981-89694981 on Build GRCh38
rs201380446 Chr.16: 89695051-89695051 on Build GRCh38
rs1025656724 Chr.16: 89695070-89695070 on Build GRCh38
rs887102804 Chr.16: 89695188-89695188 on Build GRCh38
rs867304084 Chr.16: 89695011-89695011 on Build GRCh38
rs755949658 Chr.16: 89695093-89695093 on Build GRCh38
rs937142223 Chr.16: 89695179-89695179 on Build GRCh38
rs199817238 Chr.16: 89695008-89695008 on Build GRCh38
rs754664515 Chr.16: 89695045-89695045 on Build GRCh38
rs747998262 Chr.16: 89695049-89695049 on Build GRCh38
rs751239290 Chr.16: 89695044-89695044 on Build GRCh38
rs749401269 Chr.16: 89694998-89694998 on Build GRCh38
rs752401658 Chr.16: 89695092-89695092 on Build GRCh38
rs906996831 Chr.16: 89695166-89695166 on Build GRCh38

View Full Product Details