Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94759926-94760185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf3
Entrez Gene ID
84909
Gene Name
chromosome 9 open reading frame 3
Gene Aliases
AOPEP, AP-O, APO, C90RF3, ONPEP
UniGene
Hs.434253
Gene Chromosome Location
Chr.9: 94726604-95148264 on Build GRCh38
Transcripts
NM_001193329.1 NM_001193331.2 NM_032823.5 XM_005252284.2 XM_006717306.1 XM_011519119.1 XM_011519120.2 XM_011519121.2 XM_011519122.2 XM_011519123.2 XM_011519127.1 XM_011519129.1 XM_011519130.1 XM_011519134.2 XM_017015229.1 XM_017015230.1 XM_017015231.1 XM_017015232.1 XM_017015233.1 XM_017015234.1 XM_017015235.1 XM_017015236.1 XM_017015237.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5744578 c.361C>T p.H121Y Chr.9: 94760144-94760144 on Build GRCh38
1111367 c.319A>C p.K107Q Chr.9: 94760102-94760102 on Build GRCh38
3659769 c.272C>G p.S91* Chr.9: 94760055-94760055 on Build GRCh38
1111369 c.369T>C p.S123S Chr.9: 94760152-94760152 on Build GRCh38
6616927 c.317delA p.D108fs*37 Chr.9: 94760100-94760100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778548746 Chr.9: 94760030-94760030 on Build GRCh38
rs762175674 Chr.9: 94760049-94760049 on Build GRCh38
rs777308015 Chr.9: 94760047-94760047 on Build GRCh38
rs777695268 Chr.9: 94760006-94760006 on Build GRCh38
rs371366369 Chr.9: 94760153-94760153 on Build GRCh38
rs750167553 Chr.9: 94760122-94760122 on Build GRCh38
rs139338535 Chr.9: 94760032-94760032 on Build GRCh38
rs762892735 Chr.9: 94760086-94760086 on Build GRCh38
rs202105588 Chr.9: 94760145-94760145 on Build GRCh38
rs118018100 Chr.9: 94760091-94760091 on Build GRCh38
rs144513970 Chr.9: 94760094-94760094 on Build GRCh38
rs757100689 Chr.9: 94760027-94760027 on Build GRCh38
rs770236134 Chr.9: 94760058-94760058 on Build GRCh38
rs746618229 Chr.9: 94760155-94760155 on Build GRCh38
rs779855707 Chr.9: 94760154-94760154 on Build GRCh38
rs35500870 Chr.9: 94760089-94760090 on Build GRCh38
rs767539468 Chr.9: 94760087-94760087 on Build GRCh38
rs561825699 Chr.9: 94760092-94760092 on Build GRCh38
rs199893579 Chr.9: 94760086-94760086 on Build GRCh38
rs751625506 Chr.9: 94759971-94759971 on Build GRCh38
rs765485120 Chr.9: 94760000-94760000 on Build GRCh38
rs768213104 Chr.9: 94760045-94760045 on Build GRCh38
rs746530282 Chr.9: 94760038-94760038 on Build GRCh38
rs147376186 Chr.9: 94760139-94760139 on Build GRCh38
rs1002737956 Chr.9: 94760129-94760129 on Build GRCh38
rs773445752 Chr.9: 94760063-94760063 on Build GRCh38
rs370247076 Chr.9: 94760033-94760033 on Build GRCh38
rs752252230 Chr.9: 94760076-94760076 on Build GRCh38
rs766548952 Chr.9: 94759966-94759966 on Build GRCh38
rs775795009 Chr.9: 94760026-94760027 on Build GRCh38
rs145728442 Chr.9: 94759988-94759988 on Build GRCh38
rs763295049 Chr.9: 94760074-94760074 on Build GRCh38
rs753739111 Chr.9: 94760011-94760011 on Build GRCh38

View Full Product Details