Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23614072-23614317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRICH1
Entrez Gene ID
51233
Gene Name
aspartate rich 1
Gene Aliases
C22orf43, KB-208E9.1
UniGene
Hs.517466
Gene Chromosome Location
Chr.22: 23580872-23637732 on Build GRCh38
Transcripts
NM_016449.3 XM_011530206.1 XM_011530207.2 XM_011530208.2 XM_011530209.2 XM_011530210.2 XM_017028813.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7352642 c.621C>T p.H207H Chr.22: 23614135-23614135 on Build GRCh38
3912463 c.575C>T p.S192L Chr.22: 23614181-23614181 on Build GRCh38
3552537 c.592G>A p.E198K Chr.22: 23614164-23614164 on Build GRCh38
7161120 c.598G>A p.E200K Chr.22: 23614158-23614158 on Build GRCh38
6477498 c.553G>T p.D185Y Chr.22: 23614203-23614203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757671427 Chr.22: 23614173-23614181 on Build GRCh38
rs757505826 Chr.22: 23614257-23614257 on Build GRCh38
rs969300714 Chr.22: 23614150-23614150 on Build GRCh38
rs749752277 Chr.22: 23614177-23614177 on Build GRCh38
rs115061956 Chr.22: 23614250-23614250 on Build GRCh38
rs765462624 Chr.22: 23614247-23614247 on Build GRCh38
rs773976384 Chr.22: 23614207-23614207 on Build GRCh38
rs766235140 Chr.22: 23614230-23614230 on Build GRCh38
rs760458060 Chr.22: 23614228-23614228 on Build GRCh38
rs764607178 Chr.22: 23614151-23614153 on Build GRCh38
rs553541890 Chr.22: 23614271-23614271 on Build GRCh38
rs758440426 Chr.22: 23614128-23614128 on Build GRCh38
rs965176099 Chr.22: 23614273-23614273 on Build GRCh38
rs781346908 Chr.22: 23614259-23614259 on Build GRCh38
rs767235296 Chr.22: 23614221-23614221 on Build GRCh38
rs761544147 Chr.22: 23614105-23614105 on Build GRCh38
rs777829816 Chr.22: 23614129-23614129 on Build GRCh38
rs772901935 Chr.22: 23614223-23614223 on Build GRCh38
rs370596219 Chr.22: 23614134-23614134 on Build GRCh38
rs756034068 Chr.22: 23614122-23614122 on Build GRCh38
rs753169253 Chr.22: 23614116-23614116 on Build GRCh38
rs768339400 Chr.22: 23614199-23614199 on Build GRCh38
rs761593541 Chr.22: 23614214-23614214 on Build GRCh38
rs907901509 Chr.22: 23614226-23614226 on Build GRCh38
rs999478503 Chr.22: 23614168-23614169 on Build GRCh38
rs763499359 Chr.22: 23614137-23614137 on Build GRCh38
rs183209415 Chr.22: 23614261-23614261 on Build GRCh38
rs775997324 Chr.22: 23614167-23614167 on Build GRCh38
rs74953418 Chr.22: 23614135-23614135 on Build GRCh38
rs190721080 Chr.22: 23614246-23614246 on Build GRCh38
rs763841277 Chr.22: 23614100-23614100 on Build GRCh38
rs927451927 Chr.22: 23614163-23614163 on Build GRCh38
rs117546628 Chr.22: 23614153-23614153 on Build GRCh38
rs566819064 Chr.22: 23614279-23614279 on Build GRCh38
rs759460011 Chr.22: 23614248-23614248 on Build GRCh38
rs750207560 Chr.22: 23614107-23614107 on Build GRCh38
rs528636610 Chr.22: 23614106-23614106 on Build GRCh38
rs931886933 Chr.22: 23614258-23614258 on Build GRCh38
rs766220613 Chr.22: 23614125-23614125 on Build GRCh38
rs750705049 Chr.22: 23614261-23614262 on Build GRCh38
rs1022142284 Chr.22: 23614165-23614165 on Build GRCh38
rs781222242 Chr.22: 23614259-23614260 on Build GRCh38
rs371479050 Chr.22: 23614272-23614272 on Build GRCh38
rs751854466 Chr.22: 23614150-23614151 on Build GRCh38
rs1029584209 Chr.22: 23614242-23614242 on Build GRCh38
rs766287574 Chr.22: 23614123-23614123 on Build GRCh38
rs746205514 Chr.22: 23614151-23614151 on Build GRCh38
rs375838502 Chr.22: 23614102-23614102 on Build GRCh38
rs939010592 Chr.22: 23614142-23614150 on Build GRCh38
rs899040123 Chr.22: 23614252-23614252 on Build GRCh38

View Full Product Details