Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 29862486-29862727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDIPT
Entrez Gene ID
10423
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
PIS, PIS1
UniGene
Hs.121549
Gene Chromosome Location
Chr.16: 29858356-29863288 on Build GRCh38
Transcripts
NM_001286585.1 NM_001286586.1 NM_006319.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4578920 c.86T>C p.M29T Chr.16: 29862678-29862678 on Build GRCh38
5702975 c.164G>A p.R55H Chr.16: 29862600-29862600 on Build GRCh38
380122 c.77T>C p.F26S Chr.16: 29862687-29862687 on Build GRCh38
969673 c.136C>T p.L46L Chr.16: 29862628-29862628 on Build GRCh38
969674 c.69C>A p.I23I Chr.16: 29862695-29862695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561989712 Chr.16: 29862526-29862526 on Build GRCh38
rs752899624 Chr.16: 29862672-29862673 on Build GRCh38
rs747105927 Chr.16: 29862627-29862627 on Build GRCh38
rs768801850 Chr.16: 29862643-29862643 on Build GRCh38
rs548237900 Chr.16: 29862528-29862528 on Build GRCh38
rs776673715 Chr.16: 29862645-29862645 on Build GRCh38
rs773203237 Chr.16: 29862674-29862674 on Build GRCh38
rs769650028 Chr.16: 29862657-29862657 on Build GRCh38
rs779254146 Chr.16: 29862597-29862597 on Build GRCh38
rs768753328 Chr.16: 29862621-29862621 on Build GRCh38
rs180971062 Chr.16: 29862533-29862533 on Build GRCh38
rs1013742383 Chr.16: 29862631-29862631 on Build GRCh38
rs200183429 Chr.16: 29862648-29862648 on Build GRCh38
rs1023846401 Chr.16: 29862659-29862659 on Build GRCh38
rs756387985 Chr.16: 29862693-29862695 on Build GRCh38
rs780149443 Chr.16: 29862615-29862615 on Build GRCh38
rs752418353 Chr.16: 29862554-29862554 on Build GRCh38
rs758969620 Chr.16: 29862599-29862599 on Build GRCh38
rs1033642962 Chr.16: 29862538-29862538 on Build GRCh38
rs762982857 Chr.16: 29862693-29862693 on Build GRCh38
rs367561050 Chr.16: 29862535-29862535 on Build GRCh38
rs759830426 Chr.16: 29862587-29862590 on Build GRCh38
rs184576067 Chr.16: 29862574-29862574 on Build GRCh38
rs766270100 Chr.16: 29862699-29862699 on Build GRCh38
rs202210725 Chr.16: 29862700-29862700 on Build GRCh38
rs767126124 Chr.16: 29862539-29862539 on Build GRCh38
rs746361355 Chr.16: 29862598-29862598 on Build GRCh38

View Full Product Details