Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 78248701-78248968 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP78
Entrez Gene ID
84131
Gene Name
centrosomal protein 78
Gene Aliases
C9orf81, IP63
UniGene
Hs.374421
Gene Chromosome Location
Chr.9: 78236062-78278431 on Build GRCh38
Transcripts
NM_001098802.1 NM_032171.1 XM_005252263.4 XM_005252265.2 XM_005252266.4 XM_017015192.1 XM_017015193.1 XM_017015194.1 XM_017015195.1 XM_017015196.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110336 c.966G>T p.W322C Chr.9: 78248770-78248770 on Build GRCh38
5951238 c.958-2A>G p.? Chr.9: 78248760-78248760 on Build GRCh38
5951237 c.958-2A>G p.? Chr.9: 78248760-78248760 on Build GRCh38
1110337 c.1051A>C p.K351Q Chr.9: 78248855-78248855 on Build GRCh38
6880556 c.1034G>A p.G345E Chr.9: 78248838-78248838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010645328 Chr.9: 78248911-78248911 on Build GRCh38
rs776073328 Chr.9: 78248934-78248934 on Build GRCh38
rs774484350 Chr.9: 78248792-78248792 on Build GRCh38
rs746862880 Chr.9: 78248738-78248738 on Build GRCh38
rs768400278 Chr.9: 78248817-78248817 on Build GRCh38
rs756685748 Chr.9: 78248873-78248873 on Build GRCh38
rs757318962 Chr.9: 78248906-78248906 on Build GRCh38
rs765160106 Chr.9: 78248828-78248828 on Build GRCh38
rs778203358 Chr.9: 78248894-78248894 on Build GRCh38
rs748621007 Chr.9: 78248758-78248758 on Build GRCh38
rs777933609 Chr.9: 78248786-78248786 on Build GRCh38
rs751135300 Chr.9: 78248842-78248842 on Build GRCh38
rs371728174 Chr.9: 78248847-78248847 on Build GRCh38
rs770438715 Chr.9: 78248764-78248764 on Build GRCh38
rs761677623 Chr.9: 78248826-78248826 on Build GRCh38
rs781716930 Chr.9: 78248741-78248741 on Build GRCh38
rs559801573 Chr.9: 78248791-78248791 on Build GRCh38
rs1011961824 Chr.9: 78248739-78248739 on Build GRCh38
rs374677776 Chr.9: 78248919-78248919 on Build GRCh38
rs753116427 Chr.9: 78248869-78248869 on Build GRCh38
rs762424923 Chr.9: 78248832-78248832 on Build GRCh38
rs866922612 Chr.9: 78248799-78248799 on Build GRCh38
rs373976212 Chr.9: 78248830-78248830 on Build GRCh38
rs368147059 Chr.9: 78248841-78248841 on Build GRCh38
rs751934256 Chr.9: 78248737-78248737 on Build GRCh38
rs200402728 Chr.9: 78248836-78248836 on Build GRCh38
rs776432257 Chr.9: 78248818-78248818 on Build GRCh38
rs760484787 Chr.9: 78248813-78248813 on Build GRCh38
rs201400986 Chr.9: 78248910-78248910 on Build GRCh38
rs749367291 Chr.9: 78248905-78248905 on Build GRCh38

View Full Product Details