Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74361592-74361859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD17
Entrez Gene ID
388419
Gene Name
BTB domain containing 17
Gene Aliases
BTBD17A, LGALS3BPL, TANGO10A
UniGene
Hs.211870
Gene Chromosome Location
Chr.17: 74356416-74363655 on Build GRCh38
Transcripts
NM_001080466.1 XM_011524791.2 XM_017024622.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2977448 c.21C>T p.S7S Chr.17: 74361799-74361799 on Build GRCh38
7272478 c.50C>T p.A17V Chr.17: 74361770-74361770 on Build GRCh38
5789786 c.17A>T p.Y6F Chr.17: 74361803-74361803 on Build GRCh38
4634254 c.39C>A p.G13G Chr.17: 74361781-74361781 on Build GRCh38
6277068 c.70C>A p.L24M Chr.17: 74361750-74361750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772236867 Chr.17: 74361737-74361737 on Build GRCh38
rs371360313 Chr.17: 74361662-74361662 on Build GRCh38
rs769683571 Chr.17: 74361725-74361725 on Build GRCh38
rs752443547 Chr.17: 74361835-74361835 on Build GRCh38
rs376029033 Chr.17: 74361695-74361695 on Build GRCh38
rs368356265 Chr.17: 74361830-74361830 on Build GRCh38
rs975221591 Chr.17: 74361623-74361623 on Build GRCh38
rs370346538 Chr.17: 74361838-74361838 on Build GRCh38
rs747941641 Chr.17: 74361713-74361713 on Build GRCh38
rs527274342 Chr.17: 74361674-74361674 on Build GRCh38
rs749902802 Chr.17: 74361694-74361694 on Build GRCh38
rs762339512 Chr.17: 74361687-74361687 on Build GRCh38
rs780983417 Chr.17: 74361711-74361711 on Build GRCh38
rs765815465 Chr.17: 74361811-74361811 on Build GRCh38
rs543961634 Chr.17: 74361614-74361614 on Build GRCh38
rs1022598279 Chr.17: 74361613-74361613 on Build GRCh38
rs754592789 Chr.17: 74361832-74361832 on Build GRCh38
rs757152263 Chr.17: 74361842-74361842 on Build GRCh38
rs866563989 Chr.17: 74361771-74361771 on Build GRCh38
rs887725015 Chr.17: 74361677-74361677 on Build GRCh38
rs948911150 Chr.17: 74361671-74361671 on Build GRCh38
rs576694219 Chr.17: 74361710-74361710 on Build GRCh38
rs921039818 Chr.17: 74361645-74361645 on Build GRCh38
rs912571822 Chr.17: 74361689-74361689 on Build GRCh38
rs376237249 Chr.17: 74361728-74361728 on Build GRCh38
rs201378827 Chr.17: 74361836-74361836 on Build GRCh38
rs144636871 Chr.17: 74361667-74361667 on Build GRCh38
rs12939498 Chr.17: 74361666-74361666 on Build GRCh38
rs996712270 Chr.17: 74361738-74361738 on Build GRCh38
rs765876110 Chr.17: 74361688-74361688 on Build GRCh38
rs147440277 Chr.17: 74361768-74361768 on Build GRCh38
rs1011963719 Chr.17: 74361717-74361717 on Build GRCh38
rs547419017 Chr.17: 74361729-74361729 on Build GRCh38
rs933716764 Chr.17: 74361648-74361648 on Build GRCh38
rs911229730 Chr.17: 74361665-74361665 on Build GRCh38
rs986572489 Chr.17: 74361813-74361813 on Build GRCh38
rs1013771304 Chr.17: 74361621-74361621 on Build GRCh38
rs920151936 Chr.17: 74361669-74361669 on Build GRCh38
rs762290320 Chr.17: 74361794-74361794 on Build GRCh38
rs760890251 Chr.17: 74361748-74361748 on Build GRCh38
rs1032589381 Chr.17: 74361654-74361654 on Build GRCh38
rs779539323 Chr.17: 74361696-74361696 on Build GRCh38
rs908201757 Chr.17: 74361663-74361663 on Build GRCh38
rs765973647 Chr.17: 74361829-74361829 on Build GRCh38
rs764206335 Chr.17: 74361773-74361773 on Build GRCh38
rs767111641 Chr.17: 74361834-74361834 on Build GRCh38
rs563753374 Chr.17: 74361616-74361616 on Build GRCh38
rs929165125 Chr.17: 74361670-74361670 on Build GRCh38
rs751294271 Chr.17: 74361702-74361702 on Build GRCh38
rs768761556 Chr.17: 74361754-74361754 on Build GRCh38
rs775558693 Chr.17: 74361746-74361746 on Build GRCh38
rs773894565 Chr.17: 74361812-74361812 on Build GRCh38

View Full Product Details