Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 178465555-178465824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP7
Entrez Gene ID
51661
Gene Name
FK506 binding protein 7
Gene Aliases
FKBP23, PPIase
UniGene
Hs.410378
Gene Chromosome Location
Chr.2: 178463664-178479878 on Build GRCh38
Transcripts
NM_001135212.1 NM_181342.2 XM_005246638.4 XM_011511348.2 XM_011511349.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7170015 c.624C>T p.F208F Chr.2: 178465815-178465815 on Build GRCh38
7321839 c.669G>T p.*223Y Chr.2: 178465770-178465770 on Build GRCh38
4769507 c.619G>A p.G207S Chr.2: 178465820-178465820 on Build GRCh38
5354037 c.657C>T p.H219H Chr.2: 178465782-178465782 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748099042 Chr.2: 178465724-178465724 on Build GRCh38
rs796684000 Chr.2: 178465742-178465743 on Build GRCh38
rs1003864911 Chr.2: 178465719-178465719 on Build GRCh38
rs575874003 Chr.2: 178465728-178465728 on Build GRCh38
rs962637210 Chr.2: 178465594-178465594 on Build GRCh38
rs780860193 Chr.2: 178465784-178465784 on Build GRCh38
rs777130805 Chr.2: 178465741-178465741 on Build GRCh38
rs765252154 Chr.2: 178465752-178465752 on Build GRCh38
rs1029407987 Chr.2: 178465672-178465672 on Build GRCh38
rs797014954 Chr.2: 178465752-178465752 on Build GRCh38
rs530903833 Chr.2: 178465692-178465692 on Build GRCh38
rs369967667 Chr.2: 178465781-178465781 on Build GRCh38
rs754733532 Chr.2: 178465782-178465782 on Build GRCh38
rs555612765 Chr.2: 178465653-178465653 on Build GRCh38
rs557977609 Chr.2: 178465742-178465743 on Build GRCh38
rs561585799 Chr.2: 178465791-178465791 on Build GRCh38
rs911996463 Chr.2: 178465689-178465689 on Build GRCh38
rs75348694 Chr.2: 178465751-178465752 on Build GRCh38
rs372605165 Chr.2: 178465760-178465760 on Build GRCh38
rs748784805 Chr.2: 178465795-178465795 on Build GRCh38
rs987934851 Chr.2: 178465681-178465681 on Build GRCh38
rs375380586 Chr.2: 178465774-178465774 on Build GRCh38
rs950791755 Chr.2: 178465674-178465674 on Build GRCh38
rs763063731 Chr.2: 178465769-178465769 on Build GRCh38
rs11336824 Chr.2: 178465742-178465742 on Build GRCh38
rs752627121 Chr.2: 178465787-178465787 on Build GRCh38
rs762568072 Chr.2: 178465741-178465742 on Build GRCh38
rs771266626 Chr.2: 178465642-178465642 on Build GRCh38
rs768966350 Chr.2: 178465739-178465739 on Build GRCh38
rs769520291 Chr.2: 178465740-178465741 on Build GRCh38
rs762242677 Chr.2: 178465745-178465745 on Build GRCh38

View Full Product Details