Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38309408-38309665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417281 c.947T>C p.I316T Chr.2: 38309503-38309503 on Build GRCh38
4664074 c.1020delA p.E341fs*2 Chr.2: 38309430-38309430 on Build GRCh38
6091640 c.1021G>C p.E341Q Chr.2: 38309429-38309429 on Build GRCh38
6417283 c.893T>C p.I298T Chr.2: 38309503-38309503 on Build GRCh38
4664073 c.1020delA p.E341fs*2 Chr.2: 38309430-38309430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183996817 Chr.2: 38309600-38309600 on Build GRCh38
rs192822816 Chr.2: 38309524-38309524 on Build GRCh38
rs763899562 Chr.2: 38309477-38309477 on Build GRCh38
rs770371574 Chr.2: 38309505-38309505 on Build GRCh38
rs981838190 Chr.2: 38309569-38309569 on Build GRCh38
rs558300473 Chr.2: 38309606-38309606 on Build GRCh38
rs537316002 Chr.2: 38309463-38309463 on Build GRCh38
rs145878480 Chr.2: 38309557-38309557 on Build GRCh38
rs771189954 Chr.2: 38309515-38309515 on Build GRCh38
rs961827319 Chr.2: 38309626-38309626 on Build GRCh38
rs766581581 Chr.2: 38309449-38309449 on Build GRCh38
rs757131514 Chr.2: 38309552-38309552 on Build GRCh38
rs1047422533 Chr.2: 38309532-38309532 on Build GRCh38
rs368846327 Chr.2: 38309584-38309584 on Build GRCh38
rs369343703 Chr.2: 38309539-38309539 on Build GRCh38
rs751538561 Chr.2: 38309540-38309541 on Build GRCh38
rs371444918 Chr.2: 38309450-38309450 on Build GRCh38
rs556102547 Chr.2: 38309636-38309636 on Build GRCh38
rs61754249 Chr.2: 38309485-38309485 on Build GRCh38
rs753698754 Chr.2: 38309548-38309548 on Build GRCh38
rs752639682 Chr.2: 38309468-38309468 on Build GRCh38
rs377298072 Chr.2: 38309618-38309618 on Build GRCh38
rs764060916 Chr.2: 38309545-38309545 on Build GRCh38
rs756972639 Chr.2: 38309481-38309481 on Build GRCh38
rs748678157 Chr.2: 38309494-38309494 on Build GRCh38
rs758211956 Chr.2: 38309531-38309532 on Build GRCh38
rs140725634 Chr.2: 38309537-38309537 on Build GRCh38
rs759614791 Chr.2: 38309522-38309522 on Build GRCh38
rs1012189598 Chr.2: 38309487-38309487 on Build GRCh38
rs766483956 Chr.2: 38309541-38309541 on Build GRCh38
rs773741323 Chr.2: 38309509-38309509 on Build GRCh38
rs576584865 Chr.2: 38309608-38309608 on Build GRCh38
rs941011006 Chr.2: 38309567-38309567 on Build GRCh38
rs1038150242 Chr.2: 38309579-38309579 on Build GRCh38
rs749699468 Chr.2: 38309512-38309512 on Build GRCh38
rs779638608 Chr.2: 38309490-38309490 on Build GRCh38
rs759722159 Chr.2: 38309451-38309451 on Build GRCh38
rs188539393 Chr.2: 38309635-38309635 on Build GRCh38
rs375380520 Chr.2: 38309476-38309476 on Build GRCh38
rs2305243 Chr.2: 38309484-38309484 on Build GRCh38
rs774703380 Chr.2: 38309520-38309520 on Build GRCh38

View Full Product Details