Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 462124-462395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3
Entrez Gene ID
11336
Gene Name
exocyst complex component 3
Gene Aliases
SEC6, SEC6L1, Sec6p
UniGene
Hs.481464
Gene Chromosome Location
Chr.5: 443184-471938 on Build GRCh38
Transcripts
NM_007277.4 XM_011513948.2 XM_011513949.2 XM_011513950.2 XM_011513951.2 XM_017009000.1 XM_017009001.1 XM_017009002.1 XM_017009003.1 XM_017009004.1 XM_017009005.1 XM_017009006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6011726 c.1543G>C p.E515Q Chr.5: 462197-462197 on Build GRCh38
1068518 c.1605G>A p.A535A Chr.5: 462259-462259 on Build GRCh38
7124373 c.1564C>G p.P522A Chr.5: 462218-462218 on Build GRCh38
6011727 c.1609G>C p.E537Q Chr.5: 462263-462263 on Build GRCh38
6657091 c.1653G>A p.E551E Chr.5: 462307-462307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757743642 Chr.5: 462205-462205 on Build GRCh38
rs563248749 Chr.5: 462333-462333 on Build GRCh38
rs766908267 Chr.5: 462369-462369 on Build GRCh38
rs754663864 Chr.5: 462311-462311 on Build GRCh38
rs748155022 Chr.5: 462248-462248 on Build GRCh38
rs779396819 Chr.5: 462210-462210 on Build GRCh38
rs201209420 Chr.5: 462238-462238 on Build GRCh38
rs760860067 Chr.5: 462257-462257 on Build GRCh38
rs370123001 Chr.5: 462330-462330 on Build GRCh38
rs375770333 Chr.5: 462273-462273 on Build GRCh38
rs142927762 Chr.5: 462250-462250 on Build GRCh38
rs371466267 Chr.5: 462344-462344 on Build GRCh38
rs774053027 Chr.5: 462219-462219 on Build GRCh38
rs780916912 Chr.5: 462320-462320 on Build GRCh38
rs771794323 Chr.5: 462256-462256 on Build GRCh38
rs769786148 Chr.5: 462326-462326 on Build GRCh38
rs894022430 Chr.5: 462144-462144 on Build GRCh38
rs764619730 Chr.5: 462187-462187 on Build GRCh38
rs780433692 Chr.5: 462291-462291 on Build GRCh38
rs746548341 Chr.5: 462224-462224 on Build GRCh38
rs890972 Chr.5: 462350-462350 on Build GRCh38
rs747595341 Chr.5: 462323-462323 on Build GRCh38
rs746149777 Chr.5: 462214-462214 on Build GRCh38
rs767806146 Chr.5: 462254-462254 on Build GRCh38
rs758648199 Chr.5: 462282-462282 on Build GRCh38
rs564075623 Chr.5: 462362-462362 on Build GRCh38
rs754386448 Chr.5: 462204-462204 on Build GRCh38
rs896590272 Chr.5: 462203-462203 on Build GRCh38
rs764212752 Chr.5: 462258-462258 on Build GRCh38
rs1019013793 Chr.5: 462262-462262 on Build GRCh38
rs987621686 Chr.5: 462247-462247 on Build GRCh38
rs867277355 Chr.5: 462255-462255 on Build GRCh38
rs770675110 Chr.5: 462337-462337 on Build GRCh38
rs751967905 Chr.5: 462371-462371 on Build GRCh38
rs772871065 Chr.5: 462143-462143 on Build GRCh38
rs765603186 Chr.5: 462274-462274 on Build GRCh38
rs751305470 Chr.5: 462310-462310 on Build GRCh38
rs528054276 Chr.5: 462364-462364 on Build GRCh38
rs1015616434 Chr.5: 462212-462212 on Build GRCh38
rs752411447 Chr.5: 462324-462325 on Build GRCh38
rs556077599 Chr.5: 462227-462227 on Build GRCh38
rs753906965 Chr.5: 462263-462263 on Build GRCh38
rs767569822 Chr.5: 462154-462154 on Build GRCh38
rs763494230 Chr.5: 462365-462365 on Build GRCh38
rs779522652 Chr.5: 462220-462220 on Build GRCh38
rs774114571 Chr.5: 462338-462338 on Build GRCh38
rs759871234 Chr.5: 462148-462148 on Build GRCh38
rs750819644 Chr.5: 462276-462276 on Build GRCh38
rs374263956 Chr.5: 462184-462184 on Build GRCh38
rs776701887 Chr.5: 462345-462345 on Build GRCh38
rs755535337 Chr.5: 462329-462329 on Build GRCh38
rs920594476 Chr.5: 462347-462347 on Build GRCh38
rs372707852 Chr.5: 462251-462251 on Build GRCh38

View Full Product Details