Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1799526-1799799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D4
Entrez Gene ID
93589
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 4
Gene Aliases
RCD4
UniGene
Hs.13768
Gene Chromosome Location
Chr.12: 1791957-1922606 on Build GRCh38
Transcripts
NM_172364.4 XM_011521041.2

SNP Detail

SNP ID Chromosome Location
rs558877192 Chr.12: 1799565-1799565 on Build GRCh38
rs146080972 Chr.12: 1799579-1799579 on Build GRCh38
rs376967040 Chr.12: 1799640-1799640 on Build GRCh38
rs775382992 Chr.12: 1799698-1799698 on Build GRCh38
rs762251058 Chr.12: 1799582-1799582 on Build GRCh38
rs55971855 Chr.12: 1799683-1799683 on Build GRCh38
rs867359445 Chr.12: 1799760-1799760 on Build GRCh38
rs375737327 Chr.12: 1799753-1799753 on Build GRCh38
rs760371969 Chr.12: 1799634-1799634 on Build GRCh38
rs773554589 Chr.12: 1799745-1799746 on Build GRCh38
rs754624912 Chr.12: 1799638-1799638 on Build GRCh38
rs915832658 Chr.12: 1799545-1799545 on Build GRCh38
rs746331294 Chr.12: 1799667-1799667 on Build GRCh38
rs1047992127 Chr.12: 1799591-1799591 on Build GRCh38
rs774961697 Chr.12: 1799566-1799566 on Build GRCh38
rs773453857 Chr.12: 1799631-1799631 on Build GRCh38
rs979676588 Chr.12: 1799778-1799778 on Build GRCh38
rs140079603 Chr.12: 1799627-1799627 on Build GRCh38
rs377755809 Chr.12: 1799702-1799702 on Build GRCh38
rs372494042 Chr.12: 1799632-1799632 on Build GRCh38
rs532596130 Chr.12: 1799713-1799713 on Build GRCh38
rs1009664945 Chr.12: 1799690-1799690 on Build GRCh38
rs776838903 Chr.12: 1799712-1799712 on Build GRCh38
rs1040114577 Chr.12: 1799560-1799560 on Build GRCh38
rs1044331036 Chr.12: 1799664-1799665 on Build GRCh38
rs565275719 Chr.12: 1799701-1799701 on Build GRCh38
rs746555459 Chr.12: 1799628-1799628 on Build GRCh38
rs767866079 Chr.12: 1799606-1799606 on Build GRCh38
rs771415873 Chr.12: 1799652-1799652 on Build GRCh38
rs188700805 Chr.12: 1799730-1799730 on Build GRCh38
rs774835868 Chr.12: 1799661-1799661 on Build GRCh38
rs115192664 Chr.12: 1799642-1799642 on Build GRCh38
rs868445011 Chr.12: 1799696-1799696 on Build GRCh38
rs928027073 Chr.12: 1799613-1799613 on Build GRCh38

View Full Product Details