Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2210211-2210464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRICD5
Entrez Gene ID
283870
Gene Name
BRICHOS domain containing 5
Gene Aliases
C16orf79
UniGene
Hs.647399
Gene Chromosome Location
Chr.16: 2209253-2213037 on Build GRCh38
Transcripts
NM_182563.3 XM_005255263.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040357 c.234C>G p.P78P Chr.16: 2210228-2210228 on Build GRCh38
7271382 c.195G>A p.T65T Chr.16: 2210267-2210267 on Build GRCh38
4444525 c.248T>C p.L83P Chr.16: 2210214-2210214 on Build GRCh38
5505390 c.212C>T p.P71L Chr.16: 2210250-2210250 on Build GRCh38
4151268 c.194C>T p.T65M Chr.16: 2210268-2210268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054471456 Chr.16: 2210415-2210415 on Build GRCh38
rs762977628 Chr.16: 2210241-2210242 on Build GRCh38
rs370902791 Chr.16: 2210433-2210433 on Build GRCh38
rs891484105 Chr.16: 2210414-2210414 on Build GRCh38
rs751455544 Chr.16: 2210327-2210327 on Build GRCh38
rs569721982 Chr.16: 2210377-2210377 on Build GRCh38
rs769901164 Chr.16: 2210253-2210253 on Build GRCh38
rs764938366 Chr.16: 2210231-2210231 on Build GRCh38
rs904298406 Chr.16: 2210399-2210399 on Build GRCh38
rs149312425 Chr.16: 2210268-2210268 on Build GRCh38
rs188245691 Chr.16: 2210250-2210250 on Build GRCh38
rs758240015 Chr.16: 2210237-2210237 on Build GRCh38
rs1049580804 Chr.16: 2210385-2210385 on Build GRCh38
rs376105618 Chr.16: 2210242-2210242 on Build GRCh38
rs150440520 Chr.16: 2210308-2210308 on Build GRCh38
rs746801063 Chr.16: 2210432-2210432 on Build GRCh38
rs748331645 Chr.16: 2210252-2210252 on Build GRCh38
rs1027803902 Chr.16: 2210434-2210434 on Build GRCh38
rs143561317 Chr.16: 2210423-2210423 on Build GRCh38
rs148884818 Chr.16: 2210375-2210375 on Build GRCh38
rs138206727 Chr.16: 2210364-2210364 on Build GRCh38
rs930726381 Chr.16: 2210299-2210299 on Build GRCh38
rs771456814 Chr.16: 2210261-2210261 on Build GRCh38
rs774953680 Chr.16: 2210262-2210262 on Build GRCh38
rs147431022 Chr.16: 2210271-2210271 on Build GRCh38
rs771844156 Chr.16: 2210380-2210380 on Build GRCh38
rs920709080 Chr.16: 2210283-2210283 on Build GRCh38
rs972450572 Chr.16: 2210367-2210368 on Build GRCh38
rs749645051 Chr.16: 2210258-2210258 on Build GRCh38
rs1024983964 Chr.16: 2210430-2210430 on Build GRCh38
rs965361110 Chr.16: 2210245-2210245 on Build GRCh38
rs768147839 Chr.16: 2210263-2210263 on Build GRCh38
rs141543452 Chr.16: 2210256-2210256 on Build GRCh38
rs372988245 Chr.16: 2210307-2210307 on Build GRCh38
rs866285040 Chr.16: 2210340-2210340 on Build GRCh38
rs1052823062 Chr.16: 2210404-2210404 on Build GRCh38
rs113151710 Chr.16: 2210239-2210239 on Build GRCh38
rs949807246 Chr.16: 2210393-2210393 on Build GRCh38
rs893970413 Chr.16: 2210418-2210418 on Build GRCh38
rs201146714 Chr.16: 2210232-2210232 on Build GRCh38
rs999936227 Chr.16: 2210403-2210403 on Build GRCh38
rs569568770 Chr.16: 2210277-2210277 on Build GRCh38
rs1044018131 Chr.16: 2210406-2210406 on Build GRCh38
rs764848537 Chr.16: 2210306-2210306 on Build GRCh38
rs761445097 Chr.16: 2210270-2210270 on Build GRCh38
rs907867649 Chr.16: 2210310-2210310 on Build GRCh38
rs766181728 Chr.16: 2210324-2210324 on Build GRCh38
rs1041363761 Chr.16: 2210353-2210353 on Build GRCh38
rs770768553 Chr.16: 2210435-2210435 on Build GRCh38
rs761378229 Chr.16: 2210230-2210230 on Build GRCh38
rs773202444 Chr.16: 2210381-2210381 on Build GRCh38

View Full Product Details