Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38386337-38386562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGN
Entrez Gene ID
199720
Gene Name
gametogenetin
Gene Aliases
-
UniGene
Hs.447948
Gene Chromosome Location
Chr.19: 38384347-38388603 on Build GRCh38
Transcripts
NM_152657.3 XM_005258619.4 XM_011526603.2 XM_017026451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3202258 c.856G>A p.A286T Chr.19: 38386406-38386406 on Build GRCh38
4077731 c.802C>T p.L268L Chr.19: 38386460-38386460 on Build GRCh38
4077730 c.810C>T p.G270G Chr.19: 38386452-38386452 on Build GRCh38
4077729 c.898C>T p.R300C Chr.19: 38386364-38386364 on Build GRCh38
320572 c.820G>A p.G274S Chr.19: 38386442-38386442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534052583 Chr.19: 38386494-38386494 on Build GRCh38
rs201348776 Chr.19: 38386485-38386485 on Build GRCh38
rs775402389 Chr.19: 38386433-38386433 on Build GRCh38
rs776628145 Chr.19: 38386418-38386418 on Build GRCh38
rs1016193144 Chr.19: 38386363-38386363 on Build GRCh38
rs112963255 Chr.19: 38386374-38386374 on Build GRCh38
rs767201278 Chr.19: 38386449-38386449 on Build GRCh38
rs777963751 Chr.19: 38386463-38386463 on Build GRCh38
rs747792494 Chr.19: 38386461-38386462 on Build GRCh38
rs758971805 Chr.19: 38386461-38386461 on Build GRCh38
rs141428904 Chr.19: 38386400-38386400 on Build GRCh38
rs376522804 Chr.19: 38386407-38386407 on Build GRCh38
rs755486620 Chr.19: 38386404-38386404 on Build GRCh38
rs768669148 Chr.19: 38386496-38386496 on Build GRCh38
rs772991024 Chr.19: 38386534-38386534 on Build GRCh38
rs779773806 Chr.19: 38386453-38386453 on Build GRCh38
rs539863620 Chr.19: 38386477-38386477 on Build GRCh38
rs569448235 Chr.19: 38386452-38386452 on Build GRCh38
rs751392247 Chr.19: 38386441-38386441 on Build GRCh38
rs767476334 Chr.19: 38386529-38386529 on Build GRCh38
rs771003765 Chr.19: 38386479-38386479 on Build GRCh38
rs761331213 Chr.19: 38386447-38386447 on Build GRCh38
rs764006511 Chr.19: 38386370-38386370 on Build GRCh38
rs146483189 Chr.19: 38386528-38386528 on Build GRCh38
rs763774087 Chr.19: 38386436-38386436 on Build GRCh38
rs762918506 Chr.19: 38386435-38386435 on Build GRCh38
rs756975352 Chr.19: 38386385-38386385 on Build GRCh38
rs771219197 Chr.19: 38386417-38386417 on Build GRCh38
rs753220615 Chr.19: 38386457-38386457 on Build GRCh38
rs943994547 Chr.19: 38386439-38386439 on Build GRCh38
rs200014832 Chr.19: 38386495-38386495 on Build GRCh38
rs747408047 Chr.19: 38386476-38386476 on Build GRCh38
rs772316162 Chr.19: 38386412-38386412 on Build GRCh38
rs200586670 Chr.19: 38386491-38386491 on Build GRCh38
rs910877533 Chr.19: 38386438-38386446 on Build GRCh38
rs749892638 Chr.19: 38386451-38386451 on Build GRCh38
rs368576426 Chr.19: 38386398-38386398 on Build GRCh38
rs756488758 Chr.19: 38386446-38386447 on Build GRCh38
rs759627725 Chr.19: 38386429-38386429 on Build GRCh38
rs146534768 Chr.19: 38386380-38386380 on Build GRCh38
rs778619174 Chr.19: 38386452-38386453 on Build GRCh38
rs762731681 Chr.19: 38386361-38386361 on Build GRCh38
rs929838862 Chr.19: 38386472-38386472 on Build GRCh38
rs769922028 Chr.19: 38386432-38386432 on Build GRCh38
rs773948583 Chr.19: 38386518-38386518 on Build GRCh38
rs778236481 Chr.19: 38386414-38386414 on Build GRCh38

View Full Product Details