Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153973413-153973650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L4
Entrez Gene ID
148327
Gene Name
cAMP responsive element binding protein 3 like 4
Gene Aliases
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL
UniGene
Hs.372924
Gene Chromosome Location
Chr.1: 153967679-153974364 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255979.1 NM_001255980.1 NM_001255981.1 NM_130898.3 NR_045658.1 XM_006711172.2 XM_017000372.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154581 c.924C>T p.I308I Chr.1: 153973646-153973646 on Build GRCh38
6326692 c.898-8C>T p.? Chr.1: 153973612-153973612 on Build GRCh38
6200596 c.876C>T p.A292A Chr.1: 153973443-153973443 on Build GRCh38
896479 c.901C>A p.L301I Chr.1: 153973623-153973623 on Build GRCh38
463032 c.905T>A p.L302H Chr.1: 153973627-153973627 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752859916 Chr.1: 153973580-153973580 on Build GRCh38
rs779918976 Chr.1: 153973439-153973439 on Build GRCh38
rs773546792 Chr.1: 153973492-153973492 on Build GRCh38
rs780007047 Chr.1: 153973585-153973585 on Build GRCh38
rs761631605 Chr.1: 153973582-153973582 on Build GRCh38
rs747746218 Chr.1: 153973595-153973595 on Build GRCh38
rs747713129 Chr.1: 153973542-153973542 on Build GRCh38
rs781487127 Chr.1: 153973626-153973626 on Build GRCh38
rs1033046639 Chr.1: 153973518-153973518 on Build GRCh38
rs560535374 Chr.1: 153973569-153973569 on Build GRCh38
rs750136505 Chr.1: 153973602-153973602 on Build GRCh38
rs926644875 Chr.1: 153973628-153973628 on Build GRCh38
rs201947451 Chr.1: 153973589-153973589 on Build GRCh38
rs368450287 Chr.1: 153973497-153973497 on Build GRCh38
rs778323378 Chr.1: 153973619-153973619 on Build GRCh38
rs770374896 Chr.1: 153973478-153973478 on Build GRCh38
rs189268702 Chr.1: 153973549-153973549 on Build GRCh38
rs766050808 Chr.1: 153973621-153973623 on Build GRCh38
rs780537438 Chr.1: 153973591-153973591 on Build GRCh38
rs376649545 Chr.1: 153973550-153973550 on Build GRCh38
rs765322971 Chr.1: 153973583-153973583 on Build GRCh38
rs574768837 Chr.1: 153973538-153973538 on Build GRCh38
rs747052497 Chr.1: 153973466-153973466 on Build GRCh38
rs767451151 Chr.1: 153973575-153973575 on Build GRCh38
rs762519164 Chr.1: 153973615-153973615 on Build GRCh38
rs751526004 Chr.1: 153973627-153973629 on Build GRCh38
rs180847173 Chr.1: 153973586-153973586 on Build GRCh38
rs544520413 Chr.1: 153973496-153973496 on Build GRCh38
rs767993367 Chr.1: 153973459-153973459 on Build GRCh38
rs200948716 Chr.1: 153973613-153973613 on Build GRCh38
rs141036019 Chr.1: 153973519-153973519 on Build GRCh38
rs746663081 Chr.1: 153973453-153973453 on Build GRCh38
rs143383536 Chr.1: 153973462-153973462 on Build GRCh38

View Full Product Details