Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 112649309-112649568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPZA1
Entrez Gene ID
829
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Gene Aliases
CAPPA1, CAPZ, CAZ1
UniGene
Hs.744974
Gene Chromosome Location
Chr.1: 112619453-112671619 on Build GRCh38
Transcripts
NM_006135.2 XM_011542225.2 XM_017002424.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4020728 c.104A>G p.D35G Chr.1: 112649418-112649418 on Build GRCh38
893958 c.106G>A p.V36I Chr.1: 112649420-112649420 on Build GRCh38
5377213 c.147G>A p.G49G Chr.1: 112649461-112649461 on Build GRCh38
4919512 c.139A>T p.R47W Chr.1: 112649453-112649453 on Build GRCh38
7359483 c.106G>C p.V36L Chr.1: 112649420-112649420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762120857 Chr.1: 112649389-112649389 on Build GRCh38
rs750812987 Chr.1: 112649477-112649477 on Build GRCh38
rs773154233 Chr.1: 112649400-112649400 on Build GRCh38
rs748179884 Chr.1: 112649519-112649519 on Build GRCh38
rs753793163 Chr.1: 112649420-112649420 on Build GRCh38
rs766268171 Chr.1: 112649419-112649419 on Build GRCh38
rs1047564694 Chr.1: 112649393-112649393 on Build GRCh38
rs377276486 Chr.1: 112649482-112649482 on Build GRCh38
rs1031594919 Chr.1: 112649392-112649399 on Build GRCh38
rs760507395 Chr.1: 112649407-112649407 on Build GRCh38
rs918231611 Chr.1: 112649493-112649493 on Build GRCh38
rs779136689 Chr.1: 112649513-112649513 on Build GRCh38
rs990334068 Chr.1: 112649401-112649401 on Build GRCh38
rs200661716 Chr.1: 112649491-112649491 on Build GRCh38
rs757462203 Chr.1: 112649521-112649522 on Build GRCh38
rs373380981 Chr.1: 112649509-112649509 on Build GRCh38
rs764801142 Chr.1: 112649432-112649432 on Build GRCh38
rs914813265 Chr.1: 112649402-112649402 on Build GRCh38
rs749787054 Chr.1: 112649506-112649506 on Build GRCh38
rs200308411 Chr.1: 112649469-112649469 on Build GRCh38
rs371130747 Chr.1: 112649431-112649431 on Build GRCh38
rs767781050 Chr.1: 112649395-112649395 on Build GRCh38
rs776022004 Chr.1: 112649367-112649367 on Build GRCh38
rs765805603 Chr.1: 112649408-112649410 on Build GRCh38
rs920530909 Chr.1: 112649361-112649361 on Build GRCh38
rs76726860 Chr.1: 112649445-112649445 on Build GRCh38
rs112819520 Chr.1: 112649370-112649370 on Build GRCh38
rs749365498 Chr.1: 112649368-112649368 on Build GRCh38
rs115458575 Chr.1: 112649535-112649535 on Build GRCh38
rs774493077 Chr.1: 112649374-112649374 on Build GRCh38
rs373205268 Chr.1: 112649372-112649372 on Build GRCh38

View Full Product Details