Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 31457182-31457441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD2
Entrez Gene ID
112487
Gene Name
D-tyrosyl-tRNA deacylase 2 (putative)
Gene Aliases
C14orf126
UniGene
Hs.116014
Gene Chromosome Location
Chr.14: 31446036-31457474 on Build GRCh38
Transcripts
NM_080664.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7224706 c.53T>C p.L18P Chr.14: 31457341-31457341 on Build GRCh38
3495712 c.7G>A p.E3K Chr.14: 31457387-31457387 on Build GRCh38
1246469 c.1A>G p.M1V Chr.14: 31457393-31457393 on Build GRCh38
5949040 c.48G>A p.Q16Q Chr.14: 31457346-31457346 on Build GRCh38
227107 c.16C>T p.R6W Chr.14: 31457378-31457378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761679506 Chr.14: 31457259-31457259 on Build GRCh38
rs753923115 Chr.14: 31457363-31457363 on Build GRCh38
rs947219220 Chr.14: 31457237-31457237 on Build GRCh38
rs1007027022 Chr.14: 31457226-31457226 on Build GRCh38
rs756161985 Chr.14: 31457358-31457358 on Build GRCh38
rs536985801 Chr.14: 31457251-31457251 on Build GRCh38
rs766352843 Chr.14: 31457273-31457273 on Build GRCh38
rs753906744 Chr.14: 31457421-31457421 on Build GRCh38
rs947346485 Chr.14: 31457393-31457393 on Build GRCh38
rs986015563 Chr.14: 31457289-31457289 on Build GRCh38
rs28637251 Chr.14: 31457266-31457266 on Build GRCh38
rs892934621 Chr.14: 31457230-31457230 on Build GRCh38
rs772824387 Chr.14: 31457336-31457336 on Build GRCh38
rs896863185 Chr.14: 31457262-31457262 on Build GRCh38
rs938427215 Chr.14: 31457416-31457416 on Build GRCh38
rs935293317 Chr.14: 31457407-31457407 on Build GRCh38
rs750283152 Chr.14: 31457394-31457394 on Build GRCh38
rs554496030 Chr.14: 31457225-31457225 on Build GRCh38
rs761515859 Chr.14: 31457327-31457327 on Build GRCh38
rs1052971674 Chr.14: 31457229-31457230 on Build GRCh38
rs757505742 Chr.14: 31457295-31457295 on Build GRCh38
rs776132345 Chr.14: 31457314-31457314 on Build GRCh38
rs765671297 Chr.14: 31457357-31457357 on Build GRCh38
rs750261184 Chr.14: 31457265-31457265 on Build GRCh38
rs764983961 Chr.14: 31457332-31457332 on Build GRCh38
rs376113500 Chr.14: 31457296-31457296 on Build GRCh38
rs971335888 Chr.14: 31457395-31457395 on Build GRCh38
rs761870196 Chr.14: 31457253-31457254 on Build GRCh38
rs373831117 Chr.14: 31457318-31457318 on Build GRCh38
rs752702220 Chr.14: 31457278-31457278 on Build GRCh38
rs776303827 Chr.14: 31457320-31457320 on Build GRCh38
rs536325409 Chr.14: 31457209-31457209 on Build GRCh38
rs377356312 Chr.14: 31457406-31457406 on Build GRCh38
rs374433942 Chr.14: 31457396-31457396 on Build GRCh38
rs777163239 Chr.14: 31457236-31457236 on Build GRCh38
rs757415854 Chr.14: 31457372-31457372 on Build GRCh38
rs746217178 Chr.14: 31457375-31457375 on Build GRCh38
rs756249736 Chr.14: 31457284-31457284 on Build GRCh38
rs751551766 Chr.14: 31457342-31457342 on Build GRCh38
rs765074032 Chr.14: 31457264-31457264 on Build GRCh38
rs746487215 Chr.14: 31457316-31457316 on Build GRCh38
rs774035651 Chr.14: 31457413-31457413 on Build GRCh38
rs935339968 Chr.14: 31457371-31457371 on Build GRCh38
rs914490641 Chr.14: 31457392-31457392 on Build GRCh38
rs759925286 Chr.14: 31457227-31457227 on Build GRCh38
rs17097904 Chr.14: 31457378-31457378 on Build GRCh38
rs764921013 Chr.14: 31457303-31457304 on Build GRCh38
rs776211808 Chr.14: 31457390-31457390 on Build GRCh38
rs767459476 Chr.14: 31457420-31457420 on Build GRCh38
rs542692827 Chr.14: 31457394-31457394 on Build GRCh38
rs758837659 Chr.14: 31457376-31457376 on Build GRCh38
rs1035346031 Chr.14: 31457260-31457260 on Build GRCh38
rs371573730 Chr.14: 31457362-31457362 on Build GRCh38
rs762655681 Chr.14: 31457406-31457408 on Build GRCh38
rs769316318 Chr.14: 31457388-31457388 on Build GRCh38
rs772981435 Chr.14: 31457404-31457404 on Build GRCh38
rs759404955 Chr.14: 31457419-31457419 on Build GRCh38
rs766218899 Chr.14: 31457341-31457341 on Build GRCh38
rs766242728 Chr.14: 31457412-31457412 on Build GRCh38
rs749558711 Chr.14: 31457294-31457294 on Build GRCh38
rs764595789 Chr.14: 31457207-31457207 on Build GRCh38
rs965839317 Chr.14: 31457321-31457321 on Build GRCh38
rs762863688 Chr.14: 31457340-31457340 on Build GRCh38
rs577015923 Chr.14: 31457254-31457254 on Build GRCh38
rs181408008 Chr.14: 31457244-31457244 on Build GRCh38
rs1016654925 Chr.14: 31457355-31457355 on Build GRCh38
rs116760112 Chr.14: 31457253-31457253 on Build GRCh38
rs952887831 Chr.14: 31457281-31457281 on Build GRCh38
rs772448552 Chr.14: 31457306-31457306 on Build GRCh38
rs546379159 Chr.14: 31457317-31457317 on Build GRCh38

View Full Product Details