Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 137015158-137015389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRM2
Entrez Gene ID
1129
Gene Name
cholinergic receptor muscarinic 2
Gene Aliases
HM2
UniGene
Hs.535891
Gene Chromosome Location
Chr.7: 136868652-137020255 on Build GRCh38
Transcripts
NM_000739.2 NM_001006626.1 NM_001006627.1 NM_001006628.1 NM_001006629.1 NM_001006630.1 NM_001006631.1 NM_001006632.1 XM_011515769.2 NR_046103.1

Gene Symbol
LOC349160
Entrez Gene ID
349160
Gene Name
uncharacterized LOC349160
Gene Aliases
-
UniGene
Hs.660713
Gene Chromosome Location
Chr.7: 136898773-137164341 on Build GRCh38
Transcripts
NM_000739.2 NM_001006626.1 NM_001006627.1 NM_001006628.1 NM_001006629.1 NM_001006630.1 NM_001006631.1 NM_001006632.1 XM_011515769.2 NR_046103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
374607 c.521G>A p.G174E Chr.7: 137015386-137015386 on Build GRCh38
2859414 c.461T>A p.L154H Chr.7: 137015326-137015326 on Build GRCh38
6176574 c.403_404CG>AA p.R135K Chr.7: 137015268-137015269 on Build GRCh38
3633692 c.348C>T p.I116I Chr.7: 137015213-137015213 on Build GRCh38
376314 c.304C>A p.L102M Chr.7: 137015169-137015169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536236239 Chr.7: 137015262-137015262 on Build GRCh38
rs757844531 Chr.7: 137015281-137015281 on Build GRCh38
rs951514224 Chr.7: 137015210-137015210 on Build GRCh38
rs748172206 Chr.7: 137015268-137015268 on Build GRCh38
rs142493466 Chr.7: 137015291-137015291 on Build GRCh38
rs763447557 Chr.7: 137015348-137015348 on Build GRCh38
rs754806045 Chr.7: 137015258-137015258 on Build GRCh38
rs975508576 Chr.7: 137015345-137015345 on Build GRCh38
rs776458114 Chr.7: 137015306-137015306 on Build GRCh38
rs868734955 Chr.7: 137015267-137015267 on Build GRCh38
rs777271944 Chr.7: 137015288-137015288 on Build GRCh38
rs148163637 Chr.7: 137015321-137015321 on Build GRCh38
rs751591780 Chr.7: 137015228-137015228 on Build GRCh38
rs770334609 Chr.7: 137015325-137015325 on Build GRCh38
rs757439767 Chr.7: 137015242-137015242 on Build GRCh38
rs753589267 Chr.7: 137015252-137015252 on Build GRCh38
rs375980850 Chr.7: 137015249-137015249 on Build GRCh38
rs775850694 Chr.7: 137015338-137015338 on Build GRCh38

View Full Product Details