Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70342583-70342826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COX16
Entrez Gene ID
51241
Gene Name
COX16, cytochrome c oxidase assembly homolog
Gene Aliases
C14orf112, HSPC203
UniGene
Hs.709581
Gene Chromosome Location
Chr.14: 70325081-70359731 on Build GRCh38
Transcripts
NM_001204090.1 NM_016468.6 NM_001202547.2 NM_001202548.2 NM_001202549.2

Gene Symbol
SYNJ2BP-COX16
Entrez Gene ID
100529257
Gene Name
SYNJ2BP-COX16 readthrough
Gene Aliases
-
UniGene
Hs.443661
Gene Chromosome Location
Chr.14: 70325081-70417090 on Build GRCh38
Transcripts
NM_001204090.1 NM_016468.6 NM_001202547.2 NM_001202548.2 NM_001202549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6634321 c.100C>T p.R34C Chr.14: 70342699-70342699 on Build GRCh38
4443265 c.118C>T p.R40* Chr.14: 70342681-70342681 on Build GRCh38
957369 c.70-1G>A p.? Chr.14: 70342730-70342730 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780971991 Chr.14: 70342700-70342700 on Build GRCh38
rs761014452 Chr.14: 70342681-70342681 on Build GRCh38
rs761960279 Chr.14: 70342628-70342628 on Build GRCh38
rs767719242 Chr.14: 70342647-70342647 on Build GRCh38
rs745556374 Chr.14: 70342707-70342707 on Build GRCh38
rs370035729 Chr.14: 70342762-70342762 on Build GRCh38
rs1031090618 Chr.14: 70342794-70342794 on Build GRCh38
rs200479408 Chr.14: 70342715-70342715 on Build GRCh38
rs184802547 Chr.14: 70342648-70342648 on Build GRCh38
rs750671316 Chr.14: 70342763-70342765 on Build GRCh38
rs746679453 Chr.14: 70342642-70342647 on Build GRCh38
rs769604991 Chr.14: 70342714-70342714 on Build GRCh38
rs72733767 Chr.14: 70342779-70342779 on Build GRCh38
rs889061823 Chr.14: 70342705-70342705 on Build GRCh38
rs1022200674 Chr.14: 70342695-70342695 on Build GRCh38
rs754426839 Chr.14: 70342638-70342639 on Build GRCh38
rs965815253 Chr.14: 70342769-70342769 on Build GRCh38
rs749123716 Chr.14: 70342717-70342717 on Build GRCh38
rs766766702 Chr.14: 70342659-70342659 on Build GRCh38
rs139410596 Chr.14: 70342680-70342680 on Build GRCh38
rs142141836 Chr.14: 70342729-70342729 on Build GRCh38
rs761865792 Chr.14: 70342767-70342767 on Build GRCh38
rs1048838754 Chr.14: 70342798-70342798 on Build GRCh38
rs779478939 Chr.14: 70342737-70342740 on Build GRCh38
rs761001202 Chr.14: 70342656-70342656 on Build GRCh38
rs768414282 Chr.14: 70342723-70342723 on Build GRCh38
rs754259981 Chr.14: 70342667-70342667 on Build GRCh38
rs573618970 Chr.14: 70342698-70342698 on Build GRCh38
rs549447009 Chr.14: 70342618-70342618 on Build GRCh38
rs757637967 Chr.14: 70342722-70342723 on Build GRCh38
rs774240529 Chr.14: 70342627-70342627 on Build GRCh38

View Full Product Details