Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35639875-35640124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD8
Entrez Gene ID
8325
Gene Name
frizzled class receptor 8
Gene Aliases
FZ-8, hFZ8
UniGene
Hs.302634
Gene Chromosome Location
Chr.10: 35638249-35641434 on Build GRCh38
Transcripts
NM_031866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7172003 c.1536G>A p.S512S Chr.10: 35639894-35639894 on Build GRCh38
4685987 c.1332C>T p.A444A Chr.10: 35640098-35640098 on Build GRCh38
4685986 c.1417G>A p.V473M Chr.10: 35640013-35640013 on Build GRCh38
4685985 c.1453G>A p.V485M Chr.10: 35639977-35639977 on Build GRCh38
6050264 c.1464G>A p.P488P Chr.10: 35639966-35639966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200407196 Chr.10: 35639999-35639999 on Build GRCh38
rs372099893 Chr.10: 35640100-35640100 on Build GRCh38
rs755453846 Chr.10: 35639895-35639895 on Build GRCh38
rs769988768 Chr.10: 35640095-35640095 on Build GRCh38
rs749942368 Chr.10: 35640044-35640044 on Build GRCh38
rs201736370 Chr.10: 35639959-35639959 on Build GRCh38
rs756877656 Chr.10: 35640011-35640011 on Build GRCh38
rs746988373 Chr.10: 35640061-35640061 on Build GRCh38
rs201194361 Chr.10: 35639998-35639998 on Build GRCh38
rs374758744 Chr.10: 35639983-35639983 on Build GRCh38
rs768607594 Chr.10: 35640065-35640065 on Build GRCh38
rs757901938 Chr.10: 35640048-35640048 on Build GRCh38
rs370384168 Chr.10: 35639924-35639924 on Build GRCh38
rs765395913 Chr.10: 35639969-35639969 on Build GRCh38
rs1036561388 Chr.10: 35640059-35640059 on Build GRCh38
rs931905854 Chr.10: 35639947-35639947 on Build GRCh38
rs748992936 Chr.10: 35639909-35639909 on Build GRCh38
rs367748986 Chr.10: 35639954-35639954 on Build GRCh38
rs375906360 Chr.10: 35639989-35639989 on Build GRCh38
rs373935969 Chr.10: 35639942-35639942 on Build GRCh38
rs780912212 Chr.10: 35640070-35640070 on Build GRCh38
rs747947746 Chr.10: 35640076-35640076 on Build GRCh38
rs757002438 Chr.10: 35639921-35639921 on Build GRCh38
rs773282857 Chr.10: 35639977-35639977 on Build GRCh38
rs140730812 Chr.10: 35639897-35639897 on Build GRCh38
rs753134855 Chr.10: 35640008-35640008 on Build GRCh38
rs746791027 Chr.10: 35639951-35639951 on Build GRCh38
rs368814579 Chr.10: 35640053-35640053 on Build GRCh38
rs868223182 Chr.10: 35640023-35640023 on Build GRCh38
rs371674156 Chr.10: 35639957-35639957 on Build GRCh38
rs778436404 Chr.10: 35640041-35640041 on Build GRCh38
rs771979637 Chr.10: 35639930-35639930 on Build GRCh38
rs745385936 Chr.10: 35639925-35639925 on Build GRCh38
rs752165810 Chr.10: 35639995-35639995 on Build GRCh38

View Full Product Details