Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6623075-6623300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
144658 c.8431C>T p.P2811S Chr.11: 6623245-6623245 on Build GRCh38
689872 c.8381C>T p.S2794L Chr.11: 6623295-6623295 on Build GRCh38
4924449 c.8570C>A p.P2857H Chr.11: 6623106-6623106 on Build GRCh38
5725911 c.8385C>T p.A2795A Chr.11: 6623291-6623291 on Build GRCh38
930859 c.8479C>T p.R2827W Chr.11: 6623197-6623197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757313864 Chr.11: 6623228-6623228 on Build GRCh38
rs898949918 Chr.11: 6623216-6623216 on Build GRCh38
rs530533458 Chr.11: 6623207-6623207 on Build GRCh38
rs748609148 Chr.11: 6623277-6623277 on Build GRCh38
rs917053602 Chr.11: 6623201-6623201 on Build GRCh38
rs150994160 Chr.11: 6623174-6623174 on Build GRCh38
rs768614053 Chr.11: 6623255-6623256 on Build GRCh38
rs983234768 Chr.11: 6623198-6623198 on Build GRCh38
rs35599968 Chr.11: 6623196-6623196 on Build GRCh38
rs145232866 Chr.11: 6623123-6623123 on Build GRCh38
rs747579012 Chr.11: 6623183-6623183 on Build GRCh38
rs754014876 Chr.11: 6623210-6623210 on Build GRCh38
rs201604649 Chr.11: 6623193-6623193 on Build GRCh38
rs779100406 Chr.11: 6623153-6623153 on Build GRCh38
rs750645299 Chr.11: 6623258-6623258 on Build GRCh38
rs909978688 Chr.11: 6623227-6623227 on Build GRCh38
rs915339334 Chr.11: 6623117-6623117 on Build GRCh38
rs778237141 Chr.11: 6623194-6623194 on Build GRCh38
rs139613271 Chr.11: 6623195-6623195 on Build GRCh38
rs139981407 Chr.11: 6623122-6623122 on Build GRCh38
rs767452232 Chr.11: 6623203-6623203 on Build GRCh38
rs747308612 Chr.11: 6623270-6623270 on Build GRCh38
rs759199065 Chr.11: 6623197-6623197 on Build GRCh38
rs1020034258 Chr.11: 6623107-6623107 on Build GRCh38
rs376395483 Chr.11: 6623105-6623105 on Build GRCh38
rs968056439 Chr.11: 6623143-6623143 on Build GRCh38
rs757509870 Chr.11: 6623138-6623138 on Build GRCh38
rs145646294 Chr.11: 6623233-6623233 on Build GRCh38
rs752387035 Chr.11: 6623204-6623204 on Build GRCh38
rs769276323 Chr.11: 6623271-6623271 on Build GRCh38
rs568029252 Chr.11: 6623206-6623206 on Build GRCh38
rs140214537 Chr.11: 6623259-6623259 on Build GRCh38
rs138806709 Chr.11: 6623272-6623272 on Build GRCh38
rs746286141 Chr.11: 6623173-6623173 on Build GRCh38

View Full Product Details