Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111306820-111307075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX2
Entrez Gene ID
23316
Gene Name
cut like homeobox 2
Gene Aliases
CDP2, CUTL2
UniGene
Hs.124953
Gene Chromosome Location
Chr.12: 111034024-111350556 on Build GRCh38
Transcripts
NM_015267.3 XM_011538061.2 XM_011538063.2 XM_011538069.2 XM_011538070.2 XM_017019080.1 XM_017019081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888743 c.859G>A p.D287N Chr.12: 111306921-111306921 on Build GRCh38
6638347 c.964A>C p.S322R Chr.12: 111307026-111307026 on Build GRCh38
6194665 c.951G>A p.V317V Chr.12: 111307013-111307013 on Build GRCh38
1284368 c.894G>A p.R298R Chr.12: 111306956-111306956 on Build GRCh38
4675647 c.905C>T p.A302V Chr.12: 111306967-111306967 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs931918135 Chr.12: 111306861-111306861 on Build GRCh38
rs201349032 Chr.12: 111306909-111306909 on Build GRCh38
rs1003335802 Chr.12: 111306866-111306866 on Build GRCh38
rs753137780 Chr.12: 111306995-111306995 on Build GRCh38
rs906378105 Chr.12: 111306848-111306849 on Build GRCh38
rs532339347 Chr.12: 111307041-111307041 on Build GRCh38
rs368309448 Chr.12: 111306955-111306955 on Build GRCh38
rs777252464 Chr.12: 111306905-111306905 on Build GRCh38
rs369796774 Chr.12: 111307011-111307011 on Build GRCh38
rs781531354 Chr.12: 111306876-111306876 on Build GRCh38
rs752126465 Chr.12: 111306981-111306981 on Build GRCh38
rs752521078 Chr.12: 111306873-111306874 on Build GRCh38
rs745799384 Chr.12: 111306949-111306949 on Build GRCh38
rs748882161 Chr.12: 111306999-111306999 on Build GRCh38
rs768311870 Chr.12: 111307039-111307039 on Build GRCh38
rs748393976 Chr.12: 111306877-111306877 on Build GRCh38
rs772153748 Chr.12: 111307013-111307013 on Build GRCh38
rs571934213 Chr.12: 111306867-111306867 on Build GRCh38
rs200917097 Chr.12: 111306976-111306976 on Build GRCh38
rs894576342 Chr.12: 111306895-111306895 on Build GRCh38
rs756404654 Chr.12: 111306996-111306996 on Build GRCh38
rs746857752 Chr.12: 111307020-111307020 on Build GRCh38
rs755479468 Chr.12: 111306984-111306984 on Build GRCh38
rs199740303 Chr.12: 111306954-111306954 on Build GRCh38
rs199624287 Chr.12: 111306966-111306966 on Build GRCh38
rs994997171 Chr.12: 111306967-111306967 on Build GRCh38
rs756898105 Chr.12: 111307010-111307010 on Build GRCh38
rs368058847 Chr.12: 111306997-111306997 on Build GRCh38
rs890094061 Chr.12: 111306946-111306946 on Build GRCh38
rs763158294 Chr.12: 111307055-111307055 on Build GRCh38
rs755814432 Chr.12: 111306883-111306883 on Build GRCh38
rs565046062 Chr.12: 111307017-111307017 on Build GRCh38
rs531467959 Chr.12: 111306965-111306965 on Build GRCh38
rs766431195 Chr.12: 111306974-111306974 on Build GRCh38
rs373879856 Chr.12: 111306968-111306968 on Build GRCh38
rs766521721 Chr.12: 111307056-111307056 on Build GRCh38
rs748871113 Chr.12: 111306932-111306932 on Build GRCh38
rs200854950 Chr.12: 111306947-111306947 on Build GRCh38

View Full Product Details