Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 46167372-46167629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP5
Entrez Gene ID
59084
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative)
Gene Aliases
NPP-5, NPP5
UniGene
Hs.35198
Gene Chromosome Location
Chr.6: 46159182-46171045 on Build GRCh38
Transcripts
NM_001290072.1 NM_001290073.1 NM_021572.5 XM_005249260.3 XM_011514786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205343 c.821C>A p.P274Q Chr.6: 46167442-46167442 on Build GRCh38
3874497 c.800C>A p.S267Y Chr.6: 46167463-46167463 on Build GRCh38
3745265 c.707C>T p.T236I Chr.6: 46167556-46167556 on Build GRCh38
5429657 c.725C>T p.T242M Chr.6: 46167538-46167538 on Build GRCh38
7366283 c.748A>G p.I250V Chr.6: 46167515-46167515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756281878 Chr.6: 46167428-46167428 on Build GRCh38
rs148629518 Chr.6: 46167471-46167471 on Build GRCh38
rs1042104083 Chr.6: 46167433-46167433 on Build GRCh38
rs1015568757 Chr.6: 46167594-46167594 on Build GRCh38
rs759083467 Chr.6: 46167408-46167408 on Build GRCh38
rs767552788 Chr.6: 46167567-46167567 on Build GRCh38
rs1023868303 Chr.6: 46167563-46167563 on Build GRCh38
rs866029429 Chr.6: 46167588-46167588 on Build GRCh38
rs150707319 Chr.6: 46167538-46167538 on Build GRCh38
rs768150269 Chr.6: 46167478-46167478 on Build GRCh38
rs369308300 Chr.6: 46167501-46167501 on Build GRCh38
rs1025221257 Chr.6: 46167557-46167557 on Build GRCh38
rs143412841 Chr.6: 46167494-46167494 on Build GRCh38
rs143944060 Chr.6: 46167441-46167441 on Build GRCh38
rs754888218 Chr.6: 46167553-46167553 on Build GRCh38
rs773049968 Chr.6: 46167517-46167517 on Build GRCh38
rs373053110 Chr.6: 46167537-46167537 on Build GRCh38
rs779258399 Chr.6: 46167591-46167591 on Build GRCh38
rs778039270 Chr.6: 46167434-46167434 on Build GRCh38
rs757547147 Chr.6: 46167583-46167583 on Build GRCh38
rs865931606 Chr.6: 46167445-46167445 on Build GRCh38
rs75810327 Chr.6: 46167419-46167422 on Build GRCh38
rs377355303 Chr.6: 46167579-46167579 on Build GRCh38
rs199637964 Chr.6: 46167536-46167536 on Build GRCh38
rs775989980 Chr.6: 46167481-46167481 on Build GRCh38
rs143555767 Chr.6: 46167436-46167436 on Build GRCh38
rs34450398 Chr.6: 46167546-46167546 on Build GRCh38
rs781137947 Chr.6: 46167409-46167409 on Build GRCh38
rs755045525 Chr.6: 46167395-46167395 on Build GRCh38
rs35570105 Chr.6: 46167582-46167582 on Build GRCh38
rs374908795 Chr.6: 46167497-46167497 on Build GRCh38
rs746361962 Chr.6: 46167473-46167473 on Build GRCh38
rs558953562 Chr.6: 46167413-46167416 on Build GRCh38
rs1000025781 Chr.6: 46167472-46167472 on Build GRCh38
rs970038442 Chr.6: 46167568-46167568 on Build GRCh38
rs868597398 Chr.6: 46167540-46167540 on Build GRCh38
rs752716565 Chr.6: 46167571-46167571 on Build GRCh38
rs764798008 Chr.6: 46167420-46167420 on Build GRCh38
rs748347794 Chr.6: 46167426-46167426 on Build GRCh38
rs35458749 Chr.6: 46167573-46167573 on Build GRCh38

View Full Product Details