Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 52743829-52744098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVMER34-1
Entrez Gene ID
100288413
Gene Name
endogenous retrovirus group MER34 member 1
Gene Aliases
-
UniGene
Hs.363087
Gene Chromosome Location
Chr.4: 52743517-52751640 on Build GRCh38
Transcripts
NM_001242690.1 NM_024534.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594561 c.1453G>T p.D485Y Chr.4: 52744068-52744068 on Build GRCh38
5452563 c.1670A>C p.Q557P Chr.4: 52743851-52743851 on Build GRCh38
1594563 c.1529G>A p.R510H Chr.4: 52743992-52743992 on Build GRCh38
5658872 c.1564C>T p.Q522* Chr.4: 52743957-52743957 on Build GRCh38
7335773 c.1427_1428GG>AA p.W476* Chr.4: 52744093-52744094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573078217 Chr.4: 52743993-52743993 on Build GRCh38
rs370470165 Chr.4: 52743945-52743945 on Build GRCh38
rs772069105 Chr.4: 52743878-52743878 on Build GRCh38
rs1036204795 Chr.4: 52743954-52743954 on Build GRCh38
rs769908217 Chr.4: 52743941-52743941 on Build GRCh38
rs768531057 Chr.4: 52743975-52743975 on Build GRCh38
rs956028047 Chr.4: 52743906-52743906 on Build GRCh38
rs542449779 Chr.4: 52743974-52743974 on Build GRCh38
rs61753446 Chr.4: 52743942-52743942 on Build GRCh38
rs921288904 Chr.4: 52743908-52743908 on Build GRCh38
rs1023887819 Chr.4: 52743992-52743992 on Build GRCh38
rs911829331 Chr.4: 52743946-52743946 on Build GRCh38
rs4132835 Chr.4: 52743984-52743984 on Build GRCh38
rs944566388 Chr.4: 52743953-52743953 on Build GRCh38
rs970155188 Chr.4: 52743935-52743935 on Build GRCh38
rs769662089 Chr.4: 52743983-52743983 on Build GRCh38
rs867829410 Chr.4: 52743909-52743909 on Build GRCh38
rs980205837 Chr.4: 52743955-52743955 on Build GRCh38
rs932990319 Chr.4: 52744074-52744074 on Build GRCh38
rs1035669074 Chr.4: 52743961-52743961 on Build GRCh38
rs1031557378 Chr.4: 52743871-52743871 on Build GRCh38
rs918849361 Chr.4: 52743977-52743977 on Build GRCh38
rs747052747 Chr.4: 52743879-52743879 on Build GRCh38
rs1011405729 Chr.4: 52743912-52743912 on Build GRCh38
rs999057787 Chr.4: 52744042-52744042 on Build GRCh38
rs754520855 Chr.4: 52744043-52744043 on Build GRCh38
rs901483290 Chr.4: 52744066-52744066 on Build GRCh38
rs930425978 Chr.4: 52743963-52743963 on Build GRCh38
rs1021912952 Chr.4: 52743925-52743925 on Build GRCh38
rs191885025 Chr.4: 52743940-52743940 on Build GRCh38
rs374683776 Chr.4: 52743969-52743969 on Build GRCh38
rs960043359 Chr.4: 52743970-52743970 on Build GRCh38
rs62338777 Chr.4: 52744053-52744053 on Build GRCh38

View Full Product Details