Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15508409-15508634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACE2
Entrez Gene ID
59272
Gene Name
angiotensin I converting enzyme 2
Gene Aliases
ACEH
UniGene
Hs.178098
Gene Chromosome Location
Chr.X: 15494525-15602069 on Build GRCh38
Transcripts
XM_011545551.2 NM_001320866.1 NM_001721.6 NM_203281.2 XM_017029752.1

Gene Symbol
BMX
Entrez Gene ID
660
Gene Name
BMX non-receptor tyrosine kinase
Gene Aliases
ETK, PSCTK2, PSCTK3
UniGene
Hs.495731
Gene Chromosome Location
Chr.X: 15500777-15556529 on Build GRCh38
Transcripts
XM_011545551.2 NM_001320866.1 NM_001721.6 NM_203281.2 XM_017029752.1

SNP Detail

SNP ID Chromosome Location
rs755435627 Chr.X: 15508521-15508521 on Build GRCh38
rs748699687 Chr.X: 15508605-15508605 on Build GRCh38
rs762967277 Chr.X: 15508465-15508465 on Build GRCh38
rs781394634 Chr.X: 15508506-15508506 on Build GRCh38
rs771320489 Chr.X: 15508442-15508442 on Build GRCh38
rs920632068 Chr.X: 15508578-15508578 on Build GRCh38
rs778667976 Chr.X: 15508540-15508540 on Build GRCh38
rs775195562 Chr.X: 15508512-15508512 on Build GRCh38
rs751696072 Chr.X: 15508481-15508481 on Build GRCh38
rs7878298 Chr.X: 15508598-15508598 on Build GRCh38
rs200725402 Chr.X: 15508470-15508470 on Build GRCh38
rs748829493 Chr.X: 15508526-15508526 on Build GRCh38
rs770524776 Chr.X: 15508538-15508538 on Build GRCh38
rs753311653 Chr.X: 15508511-15508511 on Build GRCh38
rs755415647 Chr.X: 15508500-15508500 on Build GRCh38
rs750510606 Chr.X: 15508461-15508461 on Build GRCh38
rs762232033 Chr.X: 15508575-15508575 on Build GRCh38

View Full Product Details