Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6451237-6451496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD27
Entrez Gene ID
939
Gene Name
CD27 molecule
Gene Aliases
S152, S152. LPFS2, T14, TNFRSF7, Tp55
UniGene
Hs.355307
Gene Chromosome Location
Chr.12: 6444867-6451718 on Build GRCh38
Transcripts
NM_001242.4 XM_011521042.2 XM_017020232.1 XM_017020233.1 XM_017020234.1 NR_015382.2

Gene Symbol
CD27-AS1
Entrez Gene ID
678655
Gene Name
CD27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655224
Gene Chromosome Location
Chr.12: 6439001-6451517 on Build GRCh38
Transcripts
NM_001242.4 XM_011521042.2 XM_017020232.1 XM_017020233.1 XM_017020234.1 NR_015382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266204 c.685G>A p.A229T Chr.12: 6451294-6451294 on Build GRCh38
6537584 c.702C>G p.Y234* Chr.12: 6451311-6451311 on Build GRCh38
4914158 c.742C>A p.Q248K Chr.12: 6451351-6451351 on Build GRCh38
6401319 c.769G>A p.A257T Chr.12: 6451378-6451378 on Build GRCh38
6367107 c.748G>A p.D250N Chr.12: 6451357-6451357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763556515 Chr.12: 6451259-6451261 on Build GRCh38
rs761400986 Chr.12: 6451432-6451432 on Build GRCh38
rs982975241 Chr.12: 6451398-6451398 on Build GRCh38
rs775402251 Chr.12: 6451421-6451421 on Build GRCh38
rs777855449 Chr.12: 6451389-6451389 on Build GRCh38
rs144240256 Chr.12: 6451294-6451294 on Build GRCh38
rs756140861 Chr.12: 6451388-6451388 on Build GRCh38
rs764932819 Chr.12: 6451440-6451440 on Build GRCh38
rs761470007 Chr.12: 6451371-6451371 on Build GRCh38
rs1003660100 Chr.12: 6451332-6451332 on Build GRCh38
rs2532502 Chr.12: 6451307-6451307 on Build GRCh38
rs373040540 Chr.12: 6451423-6451423 on Build GRCh38
rs75524088 Chr.12: 6451322-6451322 on Build GRCh38
rs1009325030 Chr.12: 6451303-6451303 on Build GRCh38
rs761504128 Chr.12: 6451389-6451390 on Build GRCh38
rs199612439 Chr.12: 6451462-6451463 on Build GRCh38
rs376329676 Chr.12: 6451321-6451321 on Build GRCh38
rs774902396 Chr.12: 6451441-6451441 on Build GRCh38
rs752125182 Chr.12: 6451284-6451284 on Build GRCh38
rs770816422 Chr.12: 6451323-6451323 on Build GRCh38
rs547361999 Chr.12: 6451457-6451457 on Build GRCh38
rs753419238 Chr.12: 6451367-6451367 on Build GRCh38
rs781758331 Chr.12: 6451385-6451385 on Build GRCh38
rs770101157 Chr.12: 6451315-6451315 on Build GRCh38
rs766655010 Chr.12: 6451372-6451372 on Build GRCh38
rs775031871 Chr.12: 6451336-6451336 on Build GRCh38
rs11569388 Chr.12: 6451463-6451464 on Build GRCh38
rs760072720 Chr.12: 6451361-6451361 on Build GRCh38
rs758686809 Chr.12: 6451259-6451259 on Build GRCh38
rs759504816 Chr.12: 6451330-6451330 on Build GRCh38
rs772039830 Chr.12: 6451414-6451414 on Build GRCh38
rs776089109 Chr.12: 6451323-6451325 on Build GRCh38
rs921153810 Chr.12: 6451467-6451467 on Build GRCh38
rs149579617 Chr.12: 6451280-6451280 on Build GRCh38
rs755561093 Chr.12: 6451378-6451378 on Build GRCh38
rs777824114 Chr.12: 6451316-6451316 on Build GRCh38
rs867460923 Chr.12: 6451283-6451283 on Build GRCh38
rs771949875 Chr.12: 6451334-6451334 on Build GRCh38
rs751930967 Chr.12: 6451375-6451375 on Build GRCh38
rs781452204 Chr.12: 6451296-6451296 on Build GRCh38
rs780668415 Chr.12: 6451274-6451274 on Build GRCh38
rs778705752 Chr.12: 6451411-6451411 on Build GRCh38
rs1003722454 Chr.12: 6451291-6451291 on Build GRCh38
rs768064484 Chr.12: 6451424-6451424 on Build GRCh38
rs372444145 Chr.12: 6451318-6451318 on Build GRCh38
rs533340002 Chr.12: 6451445-6451445 on Build GRCh38
rs943832199 Chr.12: 6451443-6451443 on Build GRCh38
rs752860747 Chr.12: 6451387-6451387 on Build GRCh38
rs34154865 Chr.12: 6451432-6451433 on Build GRCh38
rs1059501 Chr.12: 6451407-6451407 on Build GRCh38

View Full Product Details