Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43985566-43985791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT2
Entrez Gene ID
8704
Gene Name
beta-1,4-galactosyltransferase 2
Gene Aliases
B4Gal-T2, B4Gal-T3, beta4Gal-T2
UniGene
Hs.632403
Gene Chromosome Location
Chr.1: 43979202-43991171 on Build GRCh38
Transcripts
NM_001005417.2 NM_003780.4 NM_030587.2 XM_017002716.1 XM_017002717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196614 c.931C>T p.H311Y Chr.1: 43985584-43985584 on Build GRCh38
2171404 c.1027G>A p.D343N Chr.1: 43985593-43985593 on Build GRCh38
6609445 c.939C>T p.R313R Chr.1: 43985592-43985592 on Build GRCh38
6006543 c.964C>T p.Q322* Chr.1: 43985617-43985617 on Build GRCh38
2171405 c.940G>A p.D314N Chr.1: 43985593-43985593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764114218 Chr.1: 43985659-43985659 on Build GRCh38
rs533319385 Chr.1: 43985695-43985695 on Build GRCh38
rs962380114 Chr.1: 43985595-43985595 on Build GRCh38
rs369917000 Chr.1: 43985586-43985586 on Build GRCh38
rs771221434 Chr.1: 43985744-43985744 on Build GRCh38
rs373844034 Chr.1: 43985685-43985685 on Build GRCh38
rs760698451 Chr.1: 43985648-43985648 on Build GRCh38
rs747239368 Chr.1: 43985587-43985587 on Build GRCh38
rs763400893 Chr.1: 43985613-43985613 on Build GRCh38
rs1050919512 Chr.1: 43985672-43985672 on Build GRCh38
rs942457411 Chr.1: 43985701-43985701 on Build GRCh38
rs544500310 Chr.1: 43985643-43985643 on Build GRCh38
rs773719756 Chr.1: 43985605-43985605 on Build GRCh38
rs1002956689 Chr.1: 43985749-43985749 on Build GRCh38
rs11544735 Chr.1: 43985585-43985585 on Build GRCh38
rs759752050 Chr.1: 43985639-43985639 on Build GRCh38
rs112179110 Chr.1: 43985694-43985694 on Build GRCh38
rs1054641448 Chr.1: 43985704-43985704 on Build GRCh38
rs771258134 Chr.1: 43985614-43985614 on Build GRCh38
rs1041921005 Chr.1: 43985743-43985743 on Build GRCh38
rs945718631 Chr.1: 43985722-43985722 on Build GRCh38
rs779704110 Chr.1: 43985589-43985589 on Build GRCh38
rs746554871 Chr.1: 43985592-43985592 on Build GRCh38
rs929381393 Chr.1: 43985697-43985697 on Build GRCh38
rs892784158 Chr.1: 43985712-43985712 on Build GRCh38
rs775526608 Chr.1: 43985646-43985646 on Build GRCh38
rs753746890 Chr.1: 43985664-43985664 on Build GRCh38
rs770398168 Chr.1: 43985604-43985604 on Build GRCh38

View Full Product Details