Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67832144-67832399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6025762 c.233G>A p.S78N Chr.16: 67832284-67832284 on Build GRCh38
5385195 c.279C>T p.I93I Chr.16: 67832238-67832238 on Build GRCh38
4608619 c.215C>A p.S72* Chr.16: 67832302-67832302 on Build GRCh38
6626694 c.215C>T p.S72L Chr.16: 67832302-67832302 on Build GRCh38
1679192 c.236G>T p.G79V Chr.16: 67832281-67832281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762164190 Chr.16: 67832339-67832339 on Build GRCh38
rs767519219 Chr.16: 67832345-67832345 on Build GRCh38
rs746731133 Chr.16: 67832301-67832301 on Build GRCh38
rs764571606 Chr.16: 67832330-67832330 on Build GRCh38
rs778622028 Chr.16: 67832254-67832254 on Build GRCh38
rs370562425 Chr.16: 67832207-67832207 on Build GRCh38
rs764257480 Chr.16: 67832219-67832219 on Build GRCh38
rs771940819 Chr.16: 67832280-67832280 on Build GRCh38
rs746572824 Chr.16: 67832365-67832365 on Build GRCh38
rs189805146 Chr.16: 67832253-67832253 on Build GRCh38
rs751720385 Chr.16: 67832333-67832333 on Build GRCh38
rs35885250 Chr.16: 67832275-67832275 on Build GRCh38
rs751850894 Chr.16: 67832220-67832220 on Build GRCh38
rs765627364 Chr.16: 67832214-67832214 on Build GRCh38
rs748742967 Chr.16: 67832336-67832336 on Build GRCh38
rs752728935 Chr.16: 67832358-67832358 on Build GRCh38
rs757264811 Chr.16: 67832227-67832227 on Build GRCh38
rs758592509 Chr.16: 67832218-67832218 on Build GRCh38
rs758114310 Chr.16: 67832359-67832359 on Build GRCh38
rs947102331 Chr.16: 67832278-67832278 on Build GRCh38
rs908806532 Chr.16: 67832284-67832284 on Build GRCh38
rs747770807 Chr.16: 67832172-67832172 on Build GRCh38
rs113181211 Chr.16: 67832320-67832320 on Build GRCh38
rs770717022 Chr.16: 67832375-67832375 on Build GRCh38
rs753122348 Chr.16: 67832217-67832217 on Build GRCh38
rs376569561 Chr.16: 67832302-67832302 on Build GRCh38
rs760072437 Chr.16: 67832188-67832188 on Build GRCh38
rs768428967 Chr.16: 67832169-67832169 on Build GRCh38
rs760705922 Chr.16: 67832231-67832231 on Build GRCh38
rs765794727 Chr.16: 67832340-67832340 on Build GRCh38
rs747927016 Chr.16: 67832255-67832255 on Build GRCh38
rs367715523 Chr.16: 67832210-67832210 on Build GRCh38
rs781310192 Chr.16: 67832234-67832234 on Build GRCh38
rs921303526 Chr.16: 67832321-67832321 on Build GRCh38
rs772715961 Chr.16: 67832298-67832298 on Build GRCh38
rs371984432 Chr.16: 67832349-67832349 on Build GRCh38
rs535664277 Chr.16: 67832230-67832230 on Build GRCh38
rs201356704 Chr.16: 67832195-67832195 on Build GRCh38
rs772737485 Chr.16: 67832185-67832185 on Build GRCh38
rs1023746398 Chr.16: 67832192-67832192 on Build GRCh38
rs777688046 Chr.16: 67832360-67832360 on Build GRCh38
rs750516820 Chr.16: 67832347-67832347 on Build GRCh38
rs368646286 Chr.16: 67832312-67832312 on Build GRCh38
rs574119416 Chr.16: 67832171-67832171 on Build GRCh38
rs558411801 Chr.16: 67832348-67832348 on Build GRCh38
rs554029684 Chr.16: 67832251-67832251 on Build GRCh38
rs777263098 Chr.16: 67832206-67832206 on Build GRCh38
rs763200043 Chr.16: 67832323-67832323 on Build GRCh38

View Full Product Details