Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 3585747-3585972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIPC3
Entrez Gene ID
126326
Gene Name
GIPC PDZ domain containing family member 3
Gene Aliases
C19orf64, DFNB15, DFNB72, DFNB95
UniGene
Hs.266873
Gene Chromosome Location
Chr.19: 3585531-3593541 on Build GRCh38
Transcripts
NM_133261.2 XM_005259492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6388120 c.199G>T p.E67* Chr.19: 3585796-3585796 on Build GRCh38
3692646 c.223G>A p.E75K Chr.19: 3585820-3585820 on Build GRCh38
6368071 c.182T>A p.L61Q Chr.19: 3585779-3585779 on Build GRCh38
50462 c.199G>A p.E67K Chr.19: 3585796-3585796 on Build GRCh38
6386485 c.203C>A p.A68D Chr.19: 3585800-3585800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs77070566 Chr.19: 3585948-3585948 on Build GRCh38
rs749231443 Chr.19: 3585805-3585805 on Build GRCh38
rs1002952218 Chr.19: 3585842-3585842 on Build GRCh38
rs977651576 Chr.19: 3585920-3585920 on Build GRCh38
rs942647950 Chr.19: 3585771-3585771 on Build GRCh38
rs769804873 Chr.19: 3585767-3585767 on Build GRCh38
rs990413718 Chr.19: 3585898-3585898 on Build GRCh38
rs1034043210 Chr.19: 3585877-3585878 on Build GRCh38
rs1057001027 Chr.19: 3585933-3585933 on Build GRCh38
rs896476153 Chr.19: 3585947-3585947 on Build GRCh38
rs759764607 Chr.19: 3585849-3585849 on Build GRCh38
rs1021778327 Chr.19: 3585905-3585905 on Build GRCh38
rs958932086 Chr.19: 3585877-3585877 on Build GRCh38
rs967546127 Chr.19: 3585912-3585912 on Build GRCh38
rs889681720 Chr.19: 3585769-3585769 on Build GRCh38
rs906987703 Chr.19: 3585835-3585835 on Build GRCh38
rs876657466 Chr.19: 3585789-3585789 on Build GRCh38
rs770407304 Chr.19: 3585841-3585841 on Build GRCh38
rs528823465 Chr.19: 3585811-3585811 on Build GRCh38
rs774063349 Chr.19: 3585844-3585844 on Build GRCh38
rs727504653 Chr.19: 3585804-3585804 on Build GRCh38
rs993841698 Chr.19: 3585819-3585819 on Build GRCh38
rs185551765 Chr.19: 3585833-3585833 on Build GRCh38
rs565413911 Chr.19: 3585838-3585838 on Build GRCh38
rs753810022 Chr.19: 3585915-3585915 on Build GRCh38
rs876657467 Chr.19: 3585810-3585810 on Build GRCh38

View Full Product Details