Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 86812689-86812954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACH2
Entrez Gene ID
117154
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
UniGene
Hs.86603
Gene Chromosome Location
Chr.X: 86148451-86832602 on Build GRCh38
Transcripts
NM_001139514.1 NM_001139515.1 NM_053281.3 XM_011530846.2 XM_011530847.2 XM_011530848.2 XM_017029254.1 XM_017029255.1 XM_017029256.1

SNP Detail

SNP ID Chromosome Location
rs750007305 Chr.X: 86812914-86812914 on Build GRCh38
rs779986624 Chr.X: 86812916-86812916 on Build GRCh38
rs755584527 Chr.X: 86812915-86812915 on Build GRCh38
rs936511631 Chr.X: 86812771-86812771 on Build GRCh38
rs780545351 Chr.X: 86812913-86812913 on Build GRCh38
rs749246868 Chr.X: 86812925-86812925 on Build GRCh38
rs757008623 Chr.X: 86812892-86812892 on Build GRCh38
rs772992214 Chr.X: 86812932-86812932 on Build GRCh38
rs148179765 Chr.X: 86812860-86812860 on Build GRCh38
rs773394815 Chr.X: 86812823-86812823 on Build GRCh38
rs774508631 Chr.X: 86812805-86812805 on Build GRCh38
rs751215479 Chr.X: 86812883-86812883 on Build GRCh38
rs765715387 Chr.X: 86812876-86812876 on Build GRCh38
rs762056053 Chr.X: 86812807-86812807 on Build GRCh38
rs766972397 Chr.X: 86812829-86812829 on Build GRCh38
rs748352318 Chr.X: 86812933-86812933 on Build GRCh38
rs754396217 Chr.X: 86812843-86812843 on Build GRCh38
rs761076036 Chr.X: 86812828-86812828 on Build GRCh38
rs534517265 Chr.X: 86812794-86812794 on Build GRCh38
rs200206494 Chr.X: 86812812-86812812 on Build GRCh38
rs929663941 Chr.X: 86812750-86812750 on Build GRCh38
rs142010248 Chr.X: 86812904-86812904 on Build GRCh38
rs878876146 Chr.X: 86812834-86812834 on Build GRCh38
rs747511187 Chr.X: 86812847-86812847 on Build GRCh38
rs775667652 Chr.X: 86812779-86812779 on Build GRCh38

View Full Product Details