Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42117333-42117600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND4
Entrez Gene ID
389206
Gene Name
BEN domain containing 4
Gene Aliases
CCDC4
UniGene
Hs.120591
Gene Chromosome Location
Chr.4: 42110853-42152878 on Build GRCh38
Transcripts
NM_001159547.1 NM_207406.3 XM_017008185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4528529 c.1543G>A p.D515N Chr.4: 42117580-42117580 on Build GRCh38
4528450 c.1538G>A p.G513E Chr.4: 42117585-42117585 on Build GRCh38
5401214 c.1592G>A p.G531E Chr.4: 42117531-42117531 on Build GRCh38
4398193 c.1539G>A p.G513G Chr.4: 42117584-42117584 on Build GRCh38
1718153 c.1561G>A p.D521N Chr.4: 42117562-42117562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1026203540 Chr.4: 42117461-42117461 on Build GRCh38
rs978846812 Chr.4: 42117483-42117483 on Build GRCh38
rs1013897272 Chr.4: 42117485-42117485 on Build GRCh38
rs768342771 Chr.4: 42117470-42117470 on Build GRCh38
rs754408312 Chr.4: 42117527-42117527 on Build GRCh38
rs755540046 Chr.4: 42117448-42117448 on Build GRCh38
rs1001716422 Chr.4: 42117410-42117410 on Build GRCh38
rs766943011 Chr.4: 42117508-42117508 on Build GRCh38
rs115434634 Chr.4: 42117567-42117567 on Build GRCh38
rs925754871 Chr.4: 42117502-42117502 on Build GRCh38
rs774266338 Chr.4: 42117478-42117478 on Build GRCh38
rs1039153955 Chr.4: 42117368-42117368 on Build GRCh38
rs771801858 Chr.4: 42117488-42117488 on Build GRCh38
rs753156232 Chr.4: 42117540-42117540 on Build GRCh38
rs943438615 Chr.4: 42117358-42117358 on Build GRCh38
rs1005263356 Chr.4: 42117364-42117364 on Build GRCh38
rs749775468 Chr.4: 42117425-42117425 on Build GRCh38
rs913302994 Chr.4: 42117476-42117476 on Build GRCh38
rs760292852 Chr.4: 42117504-42117504 on Build GRCh38
rs772812619 Chr.4: 42117492-42117492 on Build GRCh38
rs867065665 Chr.4: 42117473-42117473 on Build GRCh38
rs535109134 Chr.4: 42117390-42117390 on Build GRCh38
rs749027109 Chr.4: 42117469-42117469 on Build GRCh38
rs780722202 Chr.4: 42117384-42117384 on Build GRCh38
rs868316270 Chr.4: 42117373-42117373 on Build GRCh38
rs369060238 Chr.4: 42117537-42117537 on Build GRCh38
rs187366202 Chr.4: 42117528-42117528 on Build GRCh38
rs960299713 Chr.4: 42117446-42117446 on Build GRCh38
rs372409870 Chr.4: 42117558-42117558 on Build GRCh38
rs779248072 Chr.4: 42117479-42117479 on Build GRCh38
rs35372996 Chr.4: 42117404-42117405 on Build GRCh38

View Full Product Details