Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143867253-143867494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPPK1
Entrez Gene ID
83481
Gene Name
epiplakin 1
Gene Aliases
EPIPL, EPIPL1
UniGene
Hs.200412
Gene Chromosome Location
Chr.8: 143857319-143879043 on Build GRCh38
Transcripts
NM_031308.3 XM_017013889.1 XM_017013890.1 XM_017013891.1 XM_017013892.1

SNP Detail

SNP ID Chromosome Location
rs782195591 Chr.8: 143867372-143867372 on Build GRCh38
rs782149613 Chr.8: 143867279-143867279 on Build GRCh38
rs949761822 Chr.8: 143867465-143867465 on Build GRCh38
rs375347181 Chr.8: 143867314-143867314 on Build GRCh38
rs377003169 Chr.8: 143867474-143867474 on Build GRCh38
rs368596970 Chr.8: 143867340-143867340 on Build GRCh38
rs781881308 Chr.8: 143867473-143867473 on Build GRCh38
rs782125978 Chr.8: 143867343-143867343 on Build GRCh38
rs561179913 Chr.8: 143867456-143867456 on Build GRCh38
rs199879536 Chr.8: 143867387-143867387 on Build GRCh38
rs782700219 Chr.8: 143867469-143867469 on Build GRCh38
rs782248529 Chr.8: 143867326-143867326 on Build GRCh38
rs1009516137 Chr.8: 143867390-143867390 on Build GRCh38
rs782406601 Chr.8: 143867320-143867320 on Build GRCh38
rs367617111 Chr.8: 143867371-143867371 on Build GRCh38
rs201495006 Chr.8: 143867361-143867361 on Build GRCh38
rs373806117 Chr.8: 143867461-143867461 on Build GRCh38
rs145468420 Chr.8: 143867431-143867431 on Build GRCh38
rs965408907 Chr.8: 143867422-143867422 on Build GRCh38
rs782648397 Chr.8: 143867368-143867368 on Build GRCh38
rs782092695 Chr.8: 143867335-143867335 on Build GRCh38
rs782694610 Chr.8: 143867322-143867322 on Build GRCh38
rs114128539 Chr.8: 143867417-143867417 on Build GRCh38
rs199828972 Chr.8: 143867280-143867280 on Build GRCh38
rs377560588 Chr.8: 143867310-143867310 on Build GRCh38
rs376560450 Chr.8: 143867449-143867449 on Build GRCh38
rs782722315 Chr.8: 143867299-143867299 on Build GRCh38
rs782210559 Chr.8: 143867440-143867440 on Build GRCh38
rs976943937 Chr.8: 143867285-143867285 on Build GRCh38
rs781792099 Chr.8: 143867349-143867349 on Build GRCh38
rs782767924 Chr.8: 143867394-143867394 on Build GRCh38
rs782661898 Chr.8: 143867424-143867424 on Build GRCh38
rs954328784 Chr.8: 143867446-143867446 on Build GRCh38
rs541086730 Chr.8: 143867442-143867442 on Build GRCh38
rs782718501 Chr.8: 143867388-143867388 on Build GRCh38
rs374869706 Chr.8: 143867434-143867434 on Build GRCh38
rs782763036 Chr.8: 143867346-143867346 on Build GRCh38
rs372536695 Chr.8: 143867291-143867291 on Build GRCh38
rs782308067 Chr.8: 143867338-143867338 on Build GRCh38
rs370847653 Chr.8: 143867311-143867311 on Build GRCh38
rs781882643 Chr.8: 143867405-143867405 on Build GRCh38
rs781931058 Chr.8: 143867339-143867339 on Build GRCh38
rs782020110 Chr.8: 143867391-143867391 on Build GRCh38
rs782560259 Chr.8: 143867308-143867308 on Build GRCh38
rs782470665 Chr.8: 143867411-143867411 on Build GRCh38
rs572259751 Chr.8: 143867445-143867445 on Build GRCh38
rs576788732 Chr.8: 143867428-143867428 on Build GRCh38
rs377058942 Chr.8: 143867296-143867296 on Build GRCh38
rs375138049 Chr.8: 143867306-143867306 on Build GRCh38
rs782200151 Chr.8: 143867316-143867316 on Build GRCh38
rs559701261 Chr.8: 143867336-143867336 on Build GRCh38
rs782460294 Chr.8: 143867418-143867418 on Build GRCh38
rs782487282 Chr.8: 143867312-143867312 on Build GRCh38
rs1044515215 Chr.8: 143867468-143867468 on Build GRCh38
rs376941147 Chr.8: 143867278-143867278 on Build GRCh38
rs554849814 Chr.8: 143867370-143867370 on Build GRCh38
rs782456492 Chr.8: 143867353-143867353 on Build GRCh38
rs569958202 Chr.8: 143867383-143867383 on Build GRCh38
rs371636220 Chr.8: 143867290-143867290 on Build GRCh38
rs371992082 Chr.8: 143867448-143867448 on Build GRCh38
rs782393717 Chr.8: 143867376-143867376 on Build GRCh38
rs369357888 Chr.8: 143867443-143867443 on Build GRCh38
rs781854550 Chr.8: 143867341-143867341 on Build GRCh38
rs782399863 Chr.8: 143867327-143867327 on Build GRCh38
rs782238212 Chr.8: 143867375-143867375 on Build GRCh38
rs781948091 Chr.8: 143867328-143867328 on Build GRCh38
rs369842241 Chr.8: 143867455-143867455 on Build GRCh38

View Full Product Details