Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 231224228-231224471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf131
Entrez Gene ID
128061
Gene Name
chromosome 1 open reading frame 131
Gene Aliases
-
UniGene
Hs.743464
Gene Chromosome Location
Chr.1: 231223763-231241187 on Build GRCh38
Transcripts
NM_001300830.1 NM_152379.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6615091 c.806G>A p.G269E Chr.1: 231224355-231224355 on Build GRCh38
5380060 c.777T>A p.I259I Chr.1: 231224384-231224384 on Build GRCh38
1340067 c.778T>G p.L260V Chr.1: 231224383-231224383 on Build GRCh38
7175786 c.775A>G p.I259V Chr.1: 231224386-231224386 on Build GRCh38
6009872 c.867C>T p.S289S Chr.1: 231224294-231224294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757661140 Chr.1: 231224277-231224277 on Build GRCh38
rs756258397 Chr.1: 231224259-231224259 on Build GRCh38
rs764411835 Chr.1: 231224268-231224268 on Build GRCh38
rs777109296 Chr.1: 231224443-231224443 on Build GRCh38
rs1056029394 Chr.1: 231224379-231224379 on Build GRCh38
rs143666971 Chr.1: 231224396-231224396 on Build GRCh38
rs201170122 Chr.1: 231224383-231224383 on Build GRCh38
rs145700786 Chr.1: 231224338-231224338 on Build GRCh38
rs948268289 Chr.1: 231224403-231224403 on Build GRCh38
rs377224537 Chr.1: 231224404-231224404 on Build GRCh38
rs369512781 Chr.1: 231224395-231224395 on Build GRCh38
rs199681084 Chr.1: 231224289-231224289 on Build GRCh38
rs748807771 Chr.1: 231224444-231224444 on Build GRCh38
rs375448320 Chr.1: 231224388-231224388 on Build GRCh38
rs759360716 Chr.1: 231224371-231224371 on Build GRCh38
rs922044072 Chr.1: 231224381-231224381 on Build GRCh38
rs562480633 Chr.1: 231224370-231224370 on Build GRCh38
rs372175061 Chr.1: 231224271-231224271 on Build GRCh38
rs769228558 Chr.1: 231224323-231224323 on Build GRCh38
rs200489074 Chr.1: 231224392-231224392 on Build GRCh38
rs777495558 Chr.1: 231224346-231224346 on Build GRCh38
rs780544365 Chr.1: 231224414-231224414 on Build GRCh38
rs1042124795 Chr.1: 231224359-231224359 on Build GRCh38
rs376699894 Chr.1: 231224256-231224256 on Build GRCh38
rs867694267 Chr.1: 231224368-231224368 on Build GRCh38
rs780530103 Chr.1: 231224304-231224304 on Build GRCh38
rs765478155 Chr.1: 231224398-231224398 on Build GRCh38
rs748845524 Chr.1: 231224348-231224348 on Build GRCh38
rs879118555 Chr.1: 231224336-231224336 on Build GRCh38
rs955391158 Chr.1: 231224247-231224247 on Build GRCh38
rs758955301 Chr.1: 231224294-231224294 on Build GRCh38
rs755646170 Chr.1: 231224440-231224440 on Build GRCh38
rs367908093 Chr.1: 231224263-231224263 on Build GRCh38
rs747726556 Chr.1: 231224316-231224316 on Build GRCh38
rs770792057 Chr.1: 231224361-231224361 on Build GRCh38
rs752881618 Chr.1: 231224250-231224250 on Build GRCh38
rs752005619 Chr.1: 231224426-231224426 on Build GRCh38
rs758761173 Chr.1: 231224412-231224412 on Build GRCh38
rs372554564 Chr.1: 231224397-231224397 on Build GRCh38
rs753945149 Chr.1: 231224273-231224273 on Build GRCh38
rs779104790 Chr.1: 231224280-231224280 on Build GRCh38

View Full Product Details