Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 130166842-130167067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX3
Entrez Gene ID
10539
Gene Name
glutaredoxin 3
Gene Aliases
GLRX4, GRX3, GRX4, PICOT, TXNL2, TXNL3
UniGene
Hs.42644
Gene Chromosome Location
Chr.10: 130136375-130182877 on Build GRCh38
Transcripts
NM_001199868.1 NM_001321980.1 NM_006541.4 XM_017015474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916204 c.670G>T p.E224* Chr.10: 130166937-130166937 on Build GRCh38
6128659 c.663A>G p.A221A Chr.10: 130166930-130166930 on Build GRCh38
6671778 c.682A>T p.I228F Chr.10: 130166949-130166949 on Build GRCh38
6671776 c.696T>C p.A232A Chr.10: 130166963-130166963 on Build GRCh38
4422174 c.672A>G p.E224E Chr.10: 130166939-130166939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751629643 Chr.10: 130167037-130167037 on Build GRCh38
rs746123750 Chr.10: 130166992-130166993 on Build GRCh38
rs771045423 Chr.10: 130166882-130166883 on Build GRCh38
rs561986638 Chr.10: 130167023-130167023 on Build GRCh38
rs138874160 Chr.10: 130166947-130166947 on Build GRCh38
rs749094930 Chr.10: 130166991-130166991 on Build GRCh38
rs1040443064 Chr.10: 130166983-130166983 on Build GRCh38
rs761618892 Chr.10: 130166904-130166904 on Build GRCh38
rs369013678 Chr.10: 130166878-130166878 on Build GRCh38
rs764322363 Chr.10: 130166974-130166974 on Build GRCh38
rs747151952 Chr.10: 130166874-130166876 on Build GRCh38
rs773886964 Chr.10: 130166902-130166902 on Build GRCh38
rs751753292 Chr.10: 130166980-130166980 on Build GRCh38
rs931996083 Chr.10: 130167028-130167028 on Build GRCh38
rs746406715 Chr.10: 130166875-130166875 on Build GRCh38
rs752892761 Chr.10: 130166956-130166956 on Build GRCh38
rs372460784 Chr.10: 130166906-130166906 on Build GRCh38
rs775115931 Chr.10: 130166882-130166882 on Build GRCh38
rs781091036 Chr.10: 130166896-130166897 on Build GRCh38
rs369417078 Chr.10: 130166895-130166895 on Build GRCh38
rs111497138 Chr.10: 130166920-130166920 on Build GRCh38
rs757580053 Chr.10: 130166996-130166996 on Build GRCh38
rs781629526 Chr.10: 130166873-130166873 on Build GRCh38
rs765398739 Chr.10: 130166953-130166953 on Build GRCh38
rs3736898 Chr.10: 130166909-130166909 on Build GRCh38
rs760738747 Chr.10: 130166949-130166949 on Build GRCh38
rs748888201 Chr.10: 130166891-130166891 on Build GRCh38
rs756679377 Chr.10: 130167030-130167030 on Build GRCh38
rs781577931 Chr.10: 130167003-130167003 on Build GRCh38
rs911221181 Chr.10: 130167043-130167043 on Build GRCh38
rs769884898 Chr.10: 130167007-130167007 on Build GRCh38

View Full Product Details