Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 103107271-103107526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L4
Entrez Gene ID
91828
Gene Name
exocyst complex component 3 like 4
Gene Aliases
C14orf73
UniGene
Hs.37712
Gene Chromosome Location
Chr.14: 103094202-103110562 on Build GRCh38
Transcripts
NM_001077594.1 XM_011537323.2 XM_011537324.2 XM_011537325.2 XM_011537327.2 XM_011537328.2 XM_011537329.2 XM_011537330.2 XM_011537331.1 XM_011537332.2 XM_011537333.2 XM_011537334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5381897 c.1667G>A p.G556D Chr.14: 103107509-103107509 on Build GRCh38
6657141 c.1620G>A p.T540T Chr.14: 103107462-103107462 on Build GRCh38
4388633 c.1672C>T p.L558F Chr.14: 103107514-103107514 on Build GRCh38
4049252 c.1584G>A p.P528P Chr.14: 103107426-103107426 on Build GRCh38
4049253 c.1624G>A p.D542N Chr.14: 103107466-103107466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376348273 Chr.14: 103107346-103107346 on Build GRCh38
rs766066245 Chr.14: 103107386-103107386 on Build GRCh38
rs927698157 Chr.14: 103107390-103107390 on Build GRCh38
rs772586533 Chr.14: 103107454-103107454 on Build GRCh38
rs372485927 Chr.14: 103107427-103107427 on Build GRCh38
rs748630355 Chr.14: 103107380-103107380 on Build GRCh38
rs975278456 Chr.14: 103107356-103107356 on Build GRCh38
rs374105891 Chr.14: 103107426-103107426 on Build GRCh38
rs760877869 Chr.14: 103107500-103107500 on Build GRCh38
rs201051646 Chr.14: 103107375-103107375 on Build GRCh38
rs199751870 Chr.14: 103107415-103107415 on Build GRCh38
rs768837492 Chr.14: 103107377-103107377 on Build GRCh38
rs764141644 Chr.14: 103107489-103107489 on Build GRCh38
rs750367530 Chr.14: 103107361-103107361 on Build GRCh38
rs755185520 Chr.14: 103107507-103107507 on Build GRCh38
rs890500724 Chr.14: 103107352-103107352 on Build GRCh38
rs568450369 Chr.14: 103107476-103107476 on Build GRCh38
rs77559438 Chr.14: 103107477-103107477 on Build GRCh38
rs369814085 Chr.14: 103107495-103107495 on Build GRCh38
rs762788433 Chr.14: 103107485-103107485 on Build GRCh38
rs549722793 Chr.14: 103107364-103107364 on Build GRCh38
rs150993709 Chr.14: 103107505-103107505 on Build GRCh38
rs773646997 Chr.14: 103107462-103107462 on Build GRCh38
rs373962199 Chr.14: 103107501-103107501 on Build GRCh38
rs779217387 Chr.14: 103107435-103107435 on Build GRCh38
rs267603884 Chr.14: 103107480-103107480 on Build GRCh38
rs376837978 Chr.14: 103107392-103107392 on Build GRCh38
rs769674896 Chr.14: 103107481-103107481 on Build GRCh38
rs772735911 Chr.14: 103107384-103107384 on Build GRCh38
rs750552954 Chr.14: 103107425-103107425 on Build GRCh38
rs977284789 Chr.14: 103107337-103107337 on Build GRCh38
rs899700878 Chr.14: 103107453-103107454 on Build GRCh38
rs368248932 Chr.14: 103107379-103107379 on Build GRCh38
rs372497346 Chr.14: 103107383-103107383 on Build GRCh38
rs1023811848 Chr.14: 103107378-103107378 on Build GRCh38
rs770339782 Chr.14: 103107466-103107466 on Build GRCh38
rs748471071 Chr.14: 103107446-103107446 on Build GRCh38
rs867367377 Chr.14: 103107348-103107348 on Build GRCh38
rs201757868 Chr.14: 103107508-103107508 on Build GRCh38
rs780090643 Chr.14: 103107374-103107374 on Build GRCh38
rs925743907 Chr.14: 103107343-103107343 on Build GRCh38
rs1039995635 Chr.14: 103107450-103107451 on Build GRCh38
rs199732363 Chr.14: 103107422-103107422 on Build GRCh38
rs775460887 Chr.14: 103107483-103107483 on Build GRCh38
rs982423852 Chr.14: 103107402-103107402 on Build GRCh38
rs533117890 Chr.14: 103107299-103107299 on Build GRCh38
rs200667781 Chr.14: 103107419-103107419 on Build GRCh38
rs967193317 Chr.14: 103107301-103107301 on Build GRCh38
rs759444867 Chr.14: 103107393-103107393 on Build GRCh38
rs764929435 Chr.14: 103107412-103107412 on Build GRCh38
rs771772859 Chr.14: 103107471-103107471 on Build GRCh38
rs201426341 Chr.14: 103107504-103107504 on Build GRCh38
rs969951691 Chr.14: 103107382-103107382 on Build GRCh38
rs909530986 Chr.14: 103107409-103107409 on Build GRCh38

View Full Product Details