Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5787825-5788074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS3L
Entrez Gene ID
56931
Gene Name
dihydrouridine synthase 3 like
Gene Aliases
DUS3
UniGene
Hs.284297
Gene Chromosome Location
Chr.19: 5785136-5791238 on Build GRCh38
Transcripts
NM_001161619.1 NM_020175.2 XM_017027020.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5508204 c.1080C>T p.D360D Chr.19: 5788039-5788039 on Build GRCh38
5643745 c.1067A>T p.H356L Chr.19: 5788052-5788052 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893244091 Chr.19: 5787871-5787871 on Build GRCh38
rs370847997 Chr.19: 5788014-5788014 on Build GRCh38
rs754535899 Chr.19: 5788044-5788044 on Build GRCh38
rs776757985 Chr.19: 5788015-5788015 on Build GRCh38
rs192445950 Chr.19: 5788021-5788021 on Build GRCh38
rs749473905 Chr.19: 5787995-5787995 on Build GRCh38
rs915687030 Chr.19: 5787855-5787855 on Build GRCh38
rs1037319166 Chr.19: 5787924-5787924 on Build GRCh38
rs753265305 Chr.19: 5788048-5788048 on Build GRCh38
rs368372219 Chr.19: 5788027-5788027 on Build GRCh38
rs761697436 Chr.19: 5788018-5788018 on Build GRCh38
rs372607593 Chr.19: 5788040-5788040 on Build GRCh38
rs575976882 Chr.19: 5787847-5787847 on Build GRCh38
rs1004868013 Chr.19: 5787898-5787898 on Build GRCh38
rs1008198899 Chr.19: 5787962-5787962 on Build GRCh38
rs572555264 Chr.19: 5787912-5787912 on Build GRCh38
rs759388019 Chr.19: 5787852-5787852 on Build GRCh38
rs148007632 Chr.19: 5788030-5788030 on Build GRCh38
rs773413995 Chr.19: 5787991-5787991 on Build GRCh38
rs12610399 Chr.19: 5787868-5787868 on Build GRCh38
rs748495273 Chr.19: 5787987-5787987 on Build GRCh38
rs1034452475 Chr.19: 5787869-5787869 on Build GRCh38
rs374593661 Chr.19: 5787982-5787982 on Build GRCh38
rs764961171 Chr.19: 5787975-5787975 on Build GRCh38
rs374680459 Chr.19: 5787953-5787953 on Build GRCh38
rs575218563 Chr.19: 5787973-5787973 on Build GRCh38
rs147553207 Chr.19: 5787848-5787848 on Build GRCh38
rs143475735 Chr.19: 5788029-5788029 on Build GRCh38
rs371542872 Chr.19: 5787979-5787979 on Build GRCh38
rs376101379 Chr.19: 5787965-5787965 on Build GRCh38
rs995697170 Chr.19: 5787930-5787930 on Build GRCh38
rs907209248 Chr.19: 5787857-5787857 on Build GRCh38
rs199504866 Chr.19: 5787978-5787978 on Build GRCh38
rs770206213 Chr.19: 5787988-5787988 on Build GRCh38
rs767407562 Chr.19: 5787856-5787856 on Build GRCh38
rs760619580 Chr.19: 5788009-5788009 on Build GRCh38
rs756401002 Chr.19: 5788050-5788050 on Build GRCh38
rs376448715 Chr.19: 5788008-5788008 on Build GRCh38
rs967025213 Chr.19: 5787927-5787927 on Build GRCh38
rs898645508 Chr.19: 5787929-5787929 on Build GRCh38
rs907410941 Chr.19: 5787895-5787895 on Build GRCh38
rs781659247 Chr.19: 5788047-5788047 on Build GRCh38
rs954123349 Chr.19: 5787955-5787955 on Build GRCh38
rs541189923 Chr.19: 5787943-5787943 on Build GRCh38
rs966775897 Chr.19: 5787972-5787972 on Build GRCh38

View Full Product Details