Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44219007-44219280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMAP1
Entrez Gene ID
55929
Gene Name
DNA methyltransferase 1 associated protein 1
Gene Aliases
DNMAP1, DNMTAP1, EAF2, MEAF2, SWC4
UniGene
Hs.8008
Gene Chromosome Location
Chr.1: 44213428-44220679 on Build GRCh38
Transcripts
NM_001034023.1 NM_001034024.1 NM_019100.4 XM_005271037.4 XM_005271039.1 XM_005271040.1 XM_006710771.3 XM_011541784.2 XM_011541785.2 XM_017001807.1 XM_017001808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203777 c.754C>T p.R252C Chr.1: 44219089-44219089 on Build GRCh38
4807970 c.848G>A p.R283Q Chr.1: 44219105-44219105 on Build GRCh38
4677954 c.782G>A p.R261Q Chr.1: 44219117-44219117 on Build GRCh38
4008364 c.851C>T p.T284M Chr.1: 44219186-44219186 on Build GRCh38
4677953 c.812A>C p.E271A Chr.1: 44219069-44219069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762471580 Chr.1: 44219054-44219054 on Build GRCh38
rs573609619 Chr.1: 44219103-44219103 on Build GRCh38
rs765869837 Chr.1: 44219196-44219196 on Build GRCh38
rs762427205 Chr.1: 44219078-44219078 on Build GRCh38
rs765633358 Chr.1: 44219079-44219079 on Build GRCh38
rs769519173 Chr.1: 44219039-44219039 on Build GRCh38
rs768560188 Chr.1: 44219028-44219028 on Build GRCh38
rs919569333 Chr.1: 44219057-44219057 on Build GRCh38
rs138578742 Chr.1: 44219116-44219116 on Build GRCh38
rs566877674 Chr.1: 44219157-44219157 on Build GRCh38
rs772994295 Chr.1: 44219041-44219041 on Build GRCh38
rs776327594 Chr.1: 44219107-44219107 on Build GRCh38
rs774115637 Chr.1: 44219181-44219181 on Build GRCh38
rs775225142 Chr.1: 44219045-44219046 on Build GRCh38
rs777381053 Chr.1: 44219169-44219169 on Build GRCh38
rs376483697 Chr.1: 44219037-44219037 on Build GRCh38
rs755107516 Chr.1: 44219156-44219156 on Build GRCh38
rs770456146 Chr.1: 44219180-44219180 on Build GRCh38
rs758501470 Chr.1: 44219104-44219104 on Build GRCh38
rs759108052 Chr.1: 44219182-44219182 on Build GRCh38
rs115178662 Chr.1: 44219064-44219064 on Build GRCh38
rs147853070 Chr.1: 44219062-44219062 on Build GRCh38
rs763416606 Chr.1: 44219223-44219223 on Build GRCh38
rs141006092 Chr.1: 44219100-44219100 on Build GRCh38
rs769752569 Chr.1: 44219165-44219165 on Build GRCh38
rs948379587 Chr.1: 44219186-44219186 on Build GRCh38
rs749027537 Chr.1: 44219255-44219255 on Build GRCh38
rs565348656 Chr.1: 44219240-44219240 on Build GRCh38
rs546986751 Chr.1: 44219115-44219115 on Build GRCh38
rs1031879757 Chr.1: 44219197-44219197 on Build GRCh38
rs750732337 Chr.1: 44219212-44219212 on Build GRCh38
rs751746563 Chr.1: 44219235-44219235 on Build GRCh38
rs149419434 Chr.1: 44219187-44219187 on Build GRCh38
rs748011064 Chr.1: 44219163-44219163 on Build GRCh38
rs766074536 Chr.1: 44219056-44219056 on Build GRCh38
rs766505833 Chr.1: 44219231-44219231 on Build GRCh38
rs1041312090 Chr.1: 44219105-44219105 on Build GRCh38
rs756020779 Chr.1: 44219250-44219250 on Build GRCh38
rs752794783 Chr.1: 44219249-44219249 on Build GRCh38
rs908463158 Chr.1: 44219091-44219091 on Build GRCh38
rs751562389 Chr.1: 44219125-44219125 on Build GRCh38
rs368497754 Chr.1: 44219070-44219070 on Build GRCh38
rs200930513 Chr.1: 44219245-44219245 on Build GRCh38
rs374686166 Chr.1: 44219254-44219254 on Build GRCh38
rs184624290 Chr.1: 44219036-44219036 on Build GRCh38
rs150290470 Chr.1: 44219183-44219183 on Build GRCh38
rs188889401 Chr.1: 44219074-44219074 on Build GRCh38
rs762372274 Chr.1: 44219192-44219192 on Build GRCh38
rs763906066 Chr.1: 44219090-44219091 on Build GRCh38
rs749154138 Chr.1: 44219179-44219179 on Build GRCh38
rs760425845 Chr.1: 44219062-44219064 on Build GRCh38

View Full Product Details