Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120970228-120970487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLI2
Entrez Gene ID
2736
Gene Name
GLI family zinc finger 2
Gene Aliases
CJS, HPE9, PHS2, THP1, THP2
UniGene
Hs.111867
Gene Chromosome Location
Chr.2: 120797291-120992653 on Build GRCh38
Transcripts
NM_005270.4 XM_006712422.2 XM_011510969.2 XM_011510970.2 XM_011510971.2 XM_011510972.2 XM_011510973.2 XM_011510974.2 XM_017003818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3565897 c.878C>T p.P293L Chr.2: 120970425-120970425 on Build GRCh38
7318554 c.909G>C p.Q303H Chr.2: 120970456-120970456 on Build GRCh38
6213910 c.866A>C p.H289P Chr.2: 120970413-120970413 on Build GRCh38
5521678 c.912G>A p.R304R Chr.2: 120970459-120970459 on Build GRCh38
1661994 c.911G>T p.R304M Chr.2: 120970458-120970458 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759191267 Chr.2: 120970357-120970357 on Build GRCh38
rs758798082 Chr.2: 120970396-120970396 on Build GRCh38
rs544836672 Chr.2: 120970275-120970275 on Build GRCh38
rs531180096 Chr.2: 120970296-120970296 on Build GRCh38
rs768416096 Chr.2: 120970450-120970450 on Build GRCh38
rs774143395 Chr.2: 120970362-120970362 on Build GRCh38
rs761750550 Chr.2: 120970455-120970455 on Build GRCh38
rs1057518696 Chr.2: 120970438-120970438 on Build GRCh38
rs927769120 Chr.2: 120970301-120970301 on Build GRCh38
rs576176718 Chr.2: 120970309-120970309 on Build GRCh38
rs759173548 Chr.2: 120970380-120970381 on Build GRCh38
rs1867906 Chr.2: 120970266-120970266 on Build GRCh38
rs755741873 Chr.2: 120970423-120970423 on Build GRCh38
rs772017200 Chr.2: 120970365-120970365 on Build GRCh38
rs974086714 Chr.2: 120970434-120970435 on Build GRCh38
rs367810027 Chr.2: 120970427-120970427 on Build GRCh38
rs765320072 Chr.2: 120970375-120970375 on Build GRCh38
rs191369989 Chr.2: 120970356-120970356 on Build GRCh38
rs371747744 Chr.2: 120970443-120970443 on Build GRCh38
rs765528898 Chr.2: 120970458-120970458 on Build GRCh38
rs1037415411 Chr.2: 120970262-120970262 on Build GRCh38
rs750821749 Chr.2: 120970386-120970386 on Build GRCh38
rs201331588 Chr.2: 120970466-120970466 on Build GRCh38
rs773277858 Chr.2: 120970460-120970460 on Build GRCh38
rs375328592 Chr.2: 120970453-120970453 on Build GRCh38
rs748678120 Chr.2: 120970437-120970437 on Build GRCh38
rs760796572 Chr.2: 120970368-120970368 on Build GRCh38
rs748956852 Chr.2: 120970353-120970353 on Build GRCh38
rs745691710 Chr.2: 120970447-120970447 on Build GRCh38
rs775363396 Chr.2: 120970366-120970366 on Build GRCh38
rs375513993 Chr.2: 120970379-120970379 on Build GRCh38
rs752088509 Chr.2: 120970416-120970416 on Build GRCh38
rs960013430 Chr.2: 120970352-120970352 on Build GRCh38
rs200853289 Chr.2: 120970418-120970418 on Build GRCh38
rs183508716 Chr.2: 120970371-120970371 on Build GRCh38
rs746947997 Chr.2: 120970448-120970448 on Build GRCh38
rs398122882 Chr.2: 120970411-120970412 on Build GRCh38
rs139360762 Chr.2: 120970370-120970370 on Build GRCh38
rs780747425 Chr.2: 120970411-120970411 on Build GRCh38

View Full Product Details