Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142864008-142864241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 NM_001280795.2 NM_004445.5 NR_104001.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs376977419 Chr.7: 142864052-142864052 on Build GRCh38
rs377536350 Chr.7: 142864098-142864098 on Build GRCh38
rs763788542 Chr.7: 142864093-142864093 on Build GRCh38
rs756659047 Chr.7: 142864108-142864108 on Build GRCh38
rs755315060 Chr.7: 142864211-142864211 on Build GRCh38
rs947996585 Chr.7: 142864205-142864205 on Build GRCh38
rs56300555 Chr.7: 142864028-142864028 on Build GRCh38
rs748259666 Chr.7: 142864053-142864053 on Build GRCh38
rs745681588 Chr.7: 142864082-142864082 on Build GRCh38
rs777875083 Chr.7: 142864219-142864219 on Build GRCh38
rs144361430 Chr.7: 142864124-142864124 on Build GRCh38
rs749562514 Chr.7: 142864044-142864044 on Build GRCh38
rs559441393 Chr.7: 142864117-142864117 on Build GRCh38
rs1051524598 Chr.7: 142864122-142864122 on Build GRCh38
rs907101747 Chr.7: 142864187-142864187 on Build GRCh38
rs201326184 Chr.7: 142864174-142864174 on Build GRCh38
rs1045338681 Chr.7: 142864206-142864206 on Build GRCh38
rs1034428477 Chr.7: 142864200-142864200 on Build GRCh38
rs868835439 Chr.7: 142864148-142864148 on Build GRCh38
rs548632943 Chr.7: 142864134-142864134 on Build GRCh38
rs769535474 Chr.7: 142864089-142864089 on Build GRCh38
rs768197335 Chr.7: 142864165-142864165 on Build GRCh38
rs773996715 Chr.7: 142864167-142864167 on Build GRCh38
rs750820390 Chr.7: 142864037-142864037 on Build GRCh38
rs956387697 Chr.7: 142864155-142864155 on Build GRCh38
rs766992201 Chr.7: 142864203-142864203 on Build GRCh38
rs768902019 Chr.7: 142864046-142864046 on Build GRCh38
rs761261838 Chr.7: 142864173-142864173 on Build GRCh38
rs368190559 Chr.7: 142864198-142864198 on Build GRCh38
rs775320635 Chr.7: 142864163-142864163 on Build GRCh38
rs889821568 Chr.7: 142864138-142864138 on Build GRCh38
rs375010348 Chr.7: 142864095-142864095 on Build GRCh38
rs541154926 Chr.7: 142864116-142864116 on Build GRCh38
rs758452768 Chr.7: 142864135-142864135 on Build GRCh38
rs536927675 Chr.7: 142864045-142864045 on Build GRCh38
rs773799287 Chr.7: 142864094-142864094 on Build GRCh38
rs201154351 Chr.7: 142864041-142864041 on Build GRCh38
rs745842622 Chr.7: 142864161-142864161 on Build GRCh38
rs8177140 Chr.7: 142864097-142864097 on Build GRCh38
rs8177173 Chr.7: 142864164-142864164 on Build GRCh38
rs780456789 Chr.7: 142864042-142864042 on Build GRCh38
rs529333505 Chr.7: 142864092-142864092 on Build GRCh38
rs142367849 Chr.7: 142864214-142864214 on Build GRCh38
rs61732974 Chr.7: 142864215-142864215 on Build GRCh38
rs570204028 Chr.7: 142864209-142864209 on Build GRCh38
rs374051998 Chr.7: 142864112-142864112 on Build GRCh38
rs772826639 Chr.7: 142864188-142864188 on Build GRCh38
rs776496596 Chr.7: 142864076-142864076 on Build GRCh38
rs200632641 Chr.7: 142864064-142864064 on Build GRCh38
rs765403799 Chr.7: 142864212-142864212 on Build GRCh38
rs370537878 Chr.7: 142864109-142864109 on Build GRCh38
rs958321807 Chr.7: 142864213-142864213 on Build GRCh38
rs533330481 Chr.7: 142864160-142864160 on Build GRCh38

View Full Product Details