Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39056304-39056577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLP1R
Entrez Gene ID
2740
Gene Name
glucagon like peptide 1 receptor
Gene Aliases
GLP-1, GLP-1-R, GLP-1R
UniGene
Hs.351883
Gene Chromosome Location
Chr.6: 39048781-39091303 on Build GRCh38
Transcripts
NM_002062.4 NR_136562.1 NR_136563.1 XM_017010750.1 XM_017010751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1078941 c.104C>T p.T35M Chr.6: 39056422-39056422 on Build GRCh38
3762052 c.119G>A p.R40Q Chr.6: 39056437-39056437 on Build GRCh38
187477 c.143G>A p.R48H Chr.6: 39056461-39056461 on Build GRCh38
6671688 c.153T>C p.T51T Chr.6: 39056471-39056471 on Build GRCh38
1208319 c.142C>T p.R48C Chr.6: 39056460-39056460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1012198223 Chr.6: 39056552-39056552 on Build GRCh38
rs769802408 Chr.6: 39056347-39056347 on Build GRCh38
rs751829967 Chr.6: 39056393-39056393 on Build GRCh38
rs796827582 Chr.6: 39056340-39056340 on Build GRCh38
rs939246929 Chr.6: 39056335-39056335 on Build GRCh38
rs761893328 Chr.6: 39056480-39056480 on Build GRCh38
rs1015189319 Chr.6: 39056342-39056342 on Build GRCh38
rs778762355 Chr.6: 39056392-39056392 on Build GRCh38
rs750122674 Chr.6: 39056485-39056485 on Build GRCh38
rs771236944 Chr.6: 39056371-39056371 on Build GRCh38
rs148158586 Chr.6: 39056422-39056422 on Build GRCh38
rs747667115 Chr.6: 39056505-39056505 on Build GRCh38
rs772175723 Chr.6: 39056369-39056369 on Build GRCh38
rs142426070 Chr.6: 39056423-39056423 on Build GRCh38
rs757920512 Chr.6: 39056538-39056538 on Build GRCh38
rs150253529 Chr.6: 39056461-39056461 on Build GRCh38
rs759907244 Chr.6: 39056376-39056376 on Build GRCh38
rs753863651 Chr.6: 39056516-39056516 on Build GRCh38
rs113225507 Chr.6: 39056553-39056553 on Build GRCh38
rs774216973 Chr.6: 39056361-39056361 on Build GRCh38
rs141990898 Chr.6: 39056436-39056436 on Build GRCh38
rs535253185 Chr.6: 39056349-39056349 on Build GRCh38
rs759948924 Chr.6: 39056453-39056453 on Build GRCh38
rs367903915 Chr.6: 39056446-39056446 on Build GRCh38
rs201343459 Chr.6: 39056536-39056536 on Build GRCh38
rs566357476 Chr.6: 39056344-39056344 on Build GRCh38
rs754994072 Chr.6: 39056521-39056521 on Build GRCh38
rs201338954 Chr.6: 39056476-39056476 on Build GRCh38
rs770196509 Chr.6: 39056460-39056460 on Build GRCh38
rs199816840 Chr.6: 39056492-39056492 on Build GRCh38
rs200066279 Chr.6: 39056432-39056432 on Build GRCh38
rs763086049 Chr.6: 39056388-39056388 on Build GRCh38
rs751670297 Chr.6: 39056478-39056478 on Build GRCh38
rs10305436 Chr.6: 39056380-39056380 on Build GRCh38
rs748823713 Chr.6: 39056357-39056357 on Build GRCh38
rs2295006 Chr.6: 39056449-39056449 on Build GRCh38
rs200116156 Chr.6: 39056332-39056332 on Build GRCh38
rs752706975 Chr.6: 39056386-39056386 on Build GRCh38
rs375821449 Chr.6: 39056415-39056415 on Build GRCh38
rs200849157 Chr.6: 39056437-39056437 on Build GRCh38
rs866312541 Chr.6: 39056410-39056410 on Build GRCh38
rs200346220 Chr.6: 39056526-39056526 on Build GRCh38
rs780129874 Chr.6: 39056503-39056503 on Build GRCh38
rs777081758 Chr.6: 39056372-39056372 on Build GRCh38
rs761096622 Chr.6: 39056365-39056365 on Build GRCh38
rs761619784 Chr.6: 39056466-39056469 on Build GRCh38
rs936101824 Chr.6: 39056527-39056527 on Build GRCh38
rs768359978 Chr.6: 39056358-39056358 on Build GRCh38
rs201231115 Chr.6: 39056455-39056455 on Build GRCh38
rs757047051 Chr.6: 39056402-39056402 on Build GRCh38
rs10305437 Chr.6: 39056462-39056462 on Build GRCh38
rs201675748 Chr.6: 39056448-39056448 on Build GRCh38
rs767797912 Chr.6: 39056483-39056483 on Build GRCh38

View Full Product Details