Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83780221-83780460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GKAP1
Entrez Gene ID
80318
Gene Name
G kinase anchoring protein 1
Gene Aliases
FKSG21, GKAP42
UniGene
Hs.522255
Gene Chromosome Location
Chr.9: 83739421-83817837 on Build GRCh38
Transcripts
NM_001135953.1 NM_025211.3 XM_005252241.2 XM_006717293.1 XM_011519058.2 XM_017015171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6428228 c.565C>T p.H189Y Chr.9: 83780402-83780402 on Build GRCh38
7290131 c.577A>G p.K193E Chr.9: 83780390-83780390 on Build GRCh38
3780145 c.563A>G p.D188G Chr.9: 83780404-83780404 on Build GRCh38
1463301 c.585G>A p.E195E Chr.9: 83780382-83780382 on Build GRCh38
5165214 c.563-5T>C p.? Chr.9: 83780409-83780409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs112907047 Chr.9: 83780284-83780284 on Build GRCh38
rs373163192 Chr.9: 83780343-83780343 on Build GRCh38
rs768310879 Chr.9: 83780413-83780414 on Build GRCh38
rs745575801 Chr.9: 83780344-83780344 on Build GRCh38
rs769173593 Chr.9: 83780388-83780388 on Build GRCh38
rs778923783 Chr.9: 83780420-83780423 on Build GRCh38
rs972443469 Chr.9: 83780252-83780252 on Build GRCh38
rs974492271 Chr.9: 83780251-83780251 on Build GRCh38
rs946375762 Chr.9: 83780315-83780315 on Build GRCh38
rs189435010 Chr.9: 83780412-83780412 on Build GRCh38
rs750500967 Chr.9: 83780415-83780415 on Build GRCh38
rs1009619848 Chr.9: 83780413-83780413 on Build GRCh38
rs927170259 Chr.9: 83780329-83780329 on Build GRCh38
rs1056266496 Chr.9: 83780419-83780419 on Build GRCh38
rs538665011 Chr.9: 83780281-83780281 on Build GRCh38
rs376311607 Chr.9: 83780367-83780367 on Build GRCh38
rs951525185 Chr.9: 83780261-83780261 on Build GRCh38
rs748833746 Chr.9: 83780332-83780332 on Build GRCh38
rs28696030 Chr.9: 83780355-83780355 on Build GRCh38
rs921853799 Chr.9: 83780310-83780310 on Build GRCh38
rs911720821 Chr.9: 83780276-83780276 on Build GRCh38
rs891718698 Chr.9: 83780368-83780368 on Build GRCh38
rs986872860 Chr.9: 83780272-83780272 on Build GRCh38
rs765364735 Chr.9: 83780394-83780394 on Build GRCh38
rs777061715 Chr.9: 83780389-83780389 on Build GRCh38
rs771732099 Chr.9: 83780358-83780358 on Build GRCh38
rs1038890123 Chr.9: 83780299-83780299 on Build GRCh38
rs918653783 Chr.9: 83780257-83780257 on Build GRCh38
rs761059174 Chr.9: 83780384-83780384 on Build GRCh38
rs770508211 Chr.9: 83780333-83780333 on Build GRCh38
rs761882263 Chr.9: 83780393-83780393 on Build GRCh38
rs751192563 Chr.9: 83780270-83780270 on Build GRCh38
rs748755928 Chr.9: 83780413-83780413 on Build GRCh38
rs935829851 Chr.9: 83780364-83780364 on Build GRCh38
rs779804701 Chr.9: 83780388-83780389 on Build GRCh38
rs867589665 Chr.9: 83780294-83780294 on Build GRCh38
rs980241113 Chr.9: 83780316-83780316 on Build GRCh38
rs575366200 Chr.9: 83780322-83780322 on Build GRCh38

View Full Product Details