Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 32722484-32722711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1L
Entrez Gene ID
10059
Gene Name
dynamin 1 like
Gene Aliases
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV
UniGene
Hs.556296
Gene Chromosome Location
Chr.12: 32679200-32745650 on Build GRCh38
Transcripts
NM_001278463.1 NM_001278464.1 NM_001278465.1 NM_001278466.1 NM_005690.4 NM_012062.4 NM_012063.3 XM_005253282.4 XM_011520543.2 XM_011520544.2 XM_017018663.1 XM_017018664.1 XM_017018665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6647555 c.1034G>A p.C345Y Chr.12: 32722588-32722588 on Build GRCh38
4575347 c.1005C>T p.Y335Y Chr.12: 32722520-32722520 on Build GRCh38
4575346 c.966C>T p.Y322Y Chr.12: 32722520-32722520 on Build GRCh38
7319229 c.998T>C p.L333S Chr.12: 32722552-32722552 on Build GRCh38
7200346 c.939T>G p.A313A Chr.12: 32722493-32722493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771883484 Chr.12: 32722509-32722509 on Build GRCh38
rs147773105 Chr.12: 32722556-32722556 on Build GRCh38
rs763293775 Chr.12: 32722588-32722588 on Build GRCh38
rs371660963 Chr.12: 32722628-32722628 on Build GRCh38
rs764487834 Chr.12: 32722605-32722605 on Build GRCh38
rs780477539 Chr.12: 32722666-32722666 on Build GRCh38
rs758981725 Chr.12: 32722543-32722543 on Build GRCh38
rs769451515 Chr.12: 32722544-32722544 on Build GRCh38
rs754082263 Chr.12: 32722669-32722669 on Build GRCh38
rs371534794 Chr.12: 32722674-32722674 on Build GRCh38
rs7137182 Chr.12: 32722643-32722643 on Build GRCh38
rs879255689 Chr.12: 32722602-32722602 on Build GRCh38
rs370882762 Chr.12: 32722538-32722538 on Build GRCh38
rs1053656884 Chr.12: 32722514-32722514 on Build GRCh38
rs144475762 Chr.12: 32722529-32722529 on Build GRCh38
rs756201432 Chr.12: 32722648-32722648 on Build GRCh38
rs118169466 Chr.12: 32722520-32722520 on Build GRCh38
rs370248038 Chr.12: 32722517-32722517 on Build GRCh38
rs374035685 Chr.12: 32722619-32722619 on Build GRCh38
rs897648599 Chr.12: 32722530-32722530 on Build GRCh38
rs746744296 Chr.12: 32722681-32722681 on Build GRCh38
rs529454614 Chr.12: 32722631-32722631 on Build GRCh38

View Full Product Details