Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35878429-35878680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL5
Entrez Gene ID
6352
Gene Name
C-C motif chemokine ligand 5
Gene Aliases
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
UniGene
Hs.514821
Gene Chromosome Location
Chr.17: 35871491-35880373 on Build GRCh38
Transcripts
NM_001278736.1 NM_002985.2

SNP Detail

SNP ID Chromosome Location
rs769462903 Chr.17: 35878482-35878482 on Build GRCh38
rs779234893 Chr.17: 35878544-35878544 on Build GRCh38
rs113918031 Chr.17: 35878585-35878585 on Build GRCh38
rs138511589 Chr.17: 35878597-35878597 on Build GRCh38
rs754918878 Chr.17: 35878586-35878586 on Build GRCh38
rs772844224 Chr.17: 35878515-35878515 on Build GRCh38
rs764470837 Chr.17: 35878555-35878555 on Build GRCh38
rs759918234 Chr.17: 35878628-35878628 on Build GRCh38
rs746642253 Chr.17: 35878605-35878605 on Build GRCh38
rs773871651 Chr.17: 35878495-35878495 on Build GRCh38
rs765912879 Chr.17: 35878632-35878632 on Build GRCh38
rs528021126 Chr.17: 35878602-35878602 on Build GRCh38
rs745726006 Chr.17: 35878480-35878480 on Build GRCh38
rs778789699 Chr.17: 35878587-35878587 on Build GRCh38
rs149044945 Chr.17: 35878572-35878572 on Build GRCh38
rs375858276 Chr.17: 35878573-35878573 on Build GRCh38
rs764274871 Chr.17: 35878648-35878648 on Build GRCh38
rs144374758 Chr.17: 35878549-35878549 on Build GRCh38
rs1014665921 Chr.17: 35878596-35878596 on Build GRCh38
rs111739476 Chr.17: 35878527-35878527 on Build GRCh38
rs749062985 Chr.17: 35878582-35878582 on Build GRCh38
rs546323250 Chr.17: 35878510-35878510 on Build GRCh38
rs144942062 Chr.17: 35878528-35878528 on Build GRCh38
rs776911285 Chr.17: 35878619-35878619 on Build GRCh38
rs749186616 Chr.17: 35878483-35878483 on Build GRCh38
rs770571198 Chr.17: 35878609-35878609 on Build GRCh38
rs763498063 Chr.17: 35878643-35878643 on Build GRCh38
rs944097088 Chr.17: 35878470-35878470 on Build GRCh38
rs935993918 Chr.17: 35878504-35878504 on Build GRCh38
rs776127483 Chr.17: 35878633-35878633 on Build GRCh38
rs368620882 Chr.17: 35878530-35878530 on Build GRCh38
rs765819517 Chr.17: 35878537-35878537 on Build GRCh38
rs778112315 Chr.17: 35878478-35878478 on Build GRCh38
rs925199061 Chr.17: 35878460-35878460 on Build GRCh38
rs772648119 Chr.17: 35878603-35878603 on Build GRCh38
rs752867204 Chr.17: 35878547-35878547 on Build GRCh38
rs766662990 Chr.17: 35878529-35878529 on Build GRCh38
rs35359791 Chr.17: 35878578-35878578 on Build GRCh38
rs759987041 Chr.17: 35878531-35878531 on Build GRCh38
rs1010340646 Chr.17: 35878525-35878526 on Build GRCh38

View Full Product Details