Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55938390-55938663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKA
Entrez Gene ID
1606
Gene Name
diacylglycerol kinase alpha
Gene Aliases
DAGK, DAGK1, DGK-alpha
UniGene
Hs.524488
Gene Chromosome Location
Chr.12: 55931076-55954023 on Build GRCh38
Transcripts
NM_001345.4 NM_201444.2 NM_201445.1 NM_201554.1 XM_005268688.2 XM_005268689.2 XM_005268690.2 XM_011537991.2 XM_011537993.2 XM_011537995.2 XM_017018900.1 XM_017018901.1 XM_017018902.1 XM_017018903.1 XM_017018904.1 XM_017018905.1 XM_017018906.1 XM_017018907.1 XM_017018908.1 XM_017018909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922900 c.393C>T p.D131D Chr.12: 55938554-55938554 on Build GRCh38
1241073 c.371C>T p.T124M Chr.12: 55938532-55938532 on Build GRCh38
4677168 c.372G>A p.T124T Chr.12: 55938533-55938533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374645352 Chr.12: 55938579-55938579 on Build GRCh38
rs758357490 Chr.12: 55938625-55938625 on Build GRCh38
rs753803878 Chr.12: 55938472-55938472 on Build GRCh38
rs144412553 Chr.12: 55938451-55938451 on Build GRCh38
rs780692770 Chr.12: 55938638-55938638 on Build GRCh38
rs371312047 Chr.12: 55938532-55938532 on Build GRCh38
rs765461498 Chr.12: 55938616-55938616 on Build GRCh38
rs752852641 Chr.12: 55938621-55938621 on Build GRCh38
rs745363675 Chr.12: 55938533-55938533 on Build GRCh38
rs765584533 Chr.12: 55938506-55938506 on Build GRCh38
rs576164263 Chr.12: 55938418-55938418 on Build GRCh38
rs1031999186 Chr.12: 55938421-55938421 on Build GRCh38
rs368802787 Chr.12: 55938465-55938465 on Build GRCh38
rs755092533 Chr.12: 55938476-55938476 on Build GRCh38
rs752744223 Chr.12: 55938486-55938486 on Build GRCh38
rs764137836 Chr.12: 55938628-55938628 on Build GRCh38
rs762348707 Chr.12: 55938574-55938574 on Build GRCh38
rs760415366 Chr.12: 55938485-55938485 on Build GRCh38
rs908166534 Chr.12: 55938603-55938603 on Build GRCh38
rs1030806319 Chr.12: 55938450-55938450 on Build GRCh38
rs766647464 Chr.12: 55938470-55938470 on Build GRCh38
rs772598958 Chr.12: 55938578-55938578 on Build GRCh38
rs766447985 Chr.12: 55938593-55938593 on Build GRCh38
rs758232701 Chr.12: 55938491-55938491 on Build GRCh38
rs750613194 Chr.12: 55938460-55938460 on Build GRCh38
rs187690093 Chr.12: 55938615-55938615 on Build GRCh38
rs202099141 Chr.12: 55938597-55938597 on Build GRCh38
rs753045610 Chr.12: 55938636-55938636 on Build GRCh38
rs771842893 Chr.12: 55938417-55938417 on Build GRCh38
rs770429616 Chr.12: 55938526-55938526 on Build GRCh38
rs774817032 Chr.12: 55938566-55938566 on Build GRCh38
rs751330011 Chr.12: 55938635-55938635 on Build GRCh38
rs773518297 Chr.12: 55938582-55938582 on Build GRCh38
rs761799538 Chr.12: 55938438-55938438 on Build GRCh38
rs1042701581 Chr.12: 55938416-55938416 on Build GRCh38
rs183271731 Chr.12: 55938484-55938484 on Build GRCh38
rs777667745 Chr.12: 55938494-55938494 on Build GRCh38
rs761116222 Chr.12: 55938592-55938592 on Build GRCh38

View Full Product Details