Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 56834954-56835217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKE
Entrez Gene ID
8526
Gene Name
diacylglycerol kinase epsilon
Gene Aliases
AHUS7, DAGK5, DAGK6, DGK, NPHS7
UniGene
Hs.239514
Gene Chromosome Location
Chr.17: 56834099-56869565 on Build GRCh38
Transcripts
NM_003647.2 XM_011525394.2 XM_011525395.2 XM_011525396.2 XM_017025243.1 XM_017025244.1 XM_017025245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
706759 c.224G>T p.C75F Chr.17: 56835019-56835019 on Build GRCh38
4269147 c.187C>G p.R63G Chr.17: 56834982-56834982 on Build GRCh38
3194610 c.254C>T p.A85V Chr.17: 56835049-56835049 on Build GRCh38
120120 c.181G>T p.G61W Chr.17: 56834976-56834976 on Build GRCh38
3519899 c.305C>T p.A102V Chr.17: 56835100-56835100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767609629 Chr.17: 56835071-56835071 on Build GRCh38
rs369611861 Chr.17: 56835107-56835107 on Build GRCh38
rs368624469 Chr.17: 56834982-56834982 on Build GRCh38
rs763932703 Chr.17: 56835187-56835187 on Build GRCh38
rs972843644 Chr.17: 56835173-56835173 on Build GRCh38
rs938904565 Chr.17: 56835171-56835171 on Build GRCh38
rs752488975 Chr.17: 56835077-56835077 on Build GRCh38
rs371967411 Chr.17: 56835032-56835032 on Build GRCh38
rs755966050 Chr.17: 56835083-56835083 on Build GRCh38
rs773920210 Chr.17: 56835038-56835038 on Build GRCh38
rs143468147 Chr.17: 56835030-56835030 on Build GRCh38
rs761370318 Chr.17: 56834975-56834975 on Build GRCh38
rs747298055 Chr.17: 56835138-56835138 on Build GRCh38
rs772683069 Chr.17: 56835149-56835149 on Build GRCh38
rs753122117 Chr.17: 56834974-56834974 on Build GRCh38
rs752681978 Chr.17: 56835180-56835180 on Build GRCh38
rs751180799 Chr.17: 56835065-56835065 on Build GRCh38
rs312262694 Chr.17: 56834983-56834983 on Build GRCh38
rs766174427 Chr.17: 56835153-56835153 on Build GRCh38
rs1023029162 Chr.17: 56835080-56835080 on Build GRCh38
rs764983358 Chr.17: 56834976-56834976 on Build GRCh38
rs759138367 Chr.17: 56835158-56835158 on Build GRCh38
rs201209316 Chr.17: 56835098-56835098 on Build GRCh38
rs781129488 Chr.17: 56835001-56835001 on Build GRCh38
rs778963327 Chr.17: 56835110-56835110 on Build GRCh38
rs755804606 Chr.17: 56835181-56835181 on Build GRCh38
rs772637651 Chr.17: 56835027-56835027 on Build GRCh38
rs769882986 Chr.17: 56835007-56835007 on Build GRCh38
rs754934356 Chr.17: 56835066-56835066 on Build GRCh38
rs1048159 Chr.17: 56834978-56834978 on Build GRCh38
rs762662843 Chr.17: 56835150-56835150 on Build GRCh38
rs201997595 Chr.17: 56835113-56835113 on Build GRCh38
rs915029037 Chr.17: 56835123-56835123 on Build GRCh38
rs747995629 Chr.17: 56835002-56835002 on Build GRCh38
rs762304023 Chr.17: 56835049-56835049 on Build GRCh38
rs200324699 Chr.17: 56835106-56835106 on Build GRCh38
rs879047850 Chr.17: 56835042-56835042 on Build GRCh38
rs564611390 Chr.17: 56835136-56835136 on Build GRCh38
rs758893686 Chr.17: 56835164-56835165 on Build GRCh38
rs755024273 Chr.17: 56834984-56834984 on Build GRCh38
rs777787526 Chr.17: 56835096-56835096 on Build GRCh38
rs749284912 Chr.17: 56835018-56835018 on Build GRCh38
rs757482556 Chr.17: 56835104-56835104 on Build GRCh38
rs772949069 Chr.17: 56835041-56835041 on Build GRCh38
rs562198537 Chr.17: 56835020-56835020 on Build GRCh38
rs551649354 Chr.17: 56835155-56835155 on Build GRCh38
rs878925112 Chr.17: 56835167-56835167 on Build GRCh38
rs148908859 Chr.17: 56835145-56835145 on Build GRCh38
rs146765015 Chr.17: 56835050-56835050 on Build GRCh38
rs775804689 Chr.17: 56835028-56835028 on Build GRCh38
rs778077040 Chr.17: 56835008-56835008 on Build GRCh38
rs969844522 Chr.17: 56835095-56835095 on Build GRCh38
rs776001127 Chr.17: 56835135-56835135 on Build GRCh38
rs745990961 Chr.17: 56835111-56835111 on Build GRCh38
rs760028073 Chr.17: 56835022-56835022 on Build GRCh38
rs769070028 Chr.17: 56835146-56835146 on Build GRCh38
rs767519633 Chr.17: 56835162-56835162 on Build GRCh38
rs998811612 Chr.17: 56834995-56834995 on Build GRCh38
rs753355975 Chr.17: 56835011-56835013 on Build GRCh38

View Full Product Details