Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176096695-176096944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM153B
Entrez Gene ID
202134
Gene Name
family with sequence similarity 153 member B
Gene Aliases
-
UniGene
Hs.368516
Gene Chromosome Location
Chr.5: 176063709-176114798 on Build GRCh38
Transcripts
NM_001265615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6880900 c.422C>T p.T141M Chr.5: 176096881-176096881 on Build GRCh38
3827711 c.430G>A p.G144S Chr.5: 176096889-176096889 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370629353 Chr.5: 176096864-176096864 on Build GRCh38
rs374136303 Chr.5: 176096854-176096854 on Build GRCh38
rs775692765 Chr.5: 176096881-176096881 on Build GRCh38
rs908759451 Chr.5: 176096844-176096844 on Build GRCh38
rs778992367 Chr.5: 176096871-176096871 on Build GRCh38
rs368317930 Chr.5: 176096855-176096855 on Build GRCh38
rs757927157 Chr.5: 176096784-176096784 on Build GRCh38
rs747979373 Chr.5: 176096799-176096799 on Build GRCh38
rs531052891 Chr.5: 176096720-176096720 on Build GRCh38
rs375185220 Chr.5: 176096876-176096876 on Build GRCh38
rs769726575 Chr.5: 176096865-176096865 on Build GRCh38
rs754914979 Chr.5: 176096863-176096863 on Build GRCh38
rs536879388 Chr.5: 176096850-176096850 on Build GRCh38
rs776608914 Chr.5: 176096798-176096798 on Build GRCh38
rs767697639 Chr.5: 176096826-176096826 on Build GRCh38
rs762105384 Chr.5: 176096891-176096891 on Build GRCh38
rs751421250 Chr.5: 176096862-176096862 on Build GRCh38
rs745326775 Chr.5: 176096775-176096775 on Build GRCh38
rs539035263 Chr.5: 176096795-176096795 on Build GRCh38
rs779795408 Chr.5: 176096786-176096786 on Build GRCh38
rs777123479 Chr.5: 176096890-176096890 on Build GRCh38
rs573718074 Chr.5: 176096856-176096856 on Build GRCh38
rs373606955 Chr.5: 176096773-176096773 on Build GRCh38
rs370394492 Chr.5: 176096833-176096833 on Build GRCh38
rs754149094 Chr.5: 176096836-176096836 on Build GRCh38
rs561450039 Chr.5: 176096746-176096746 on Build GRCh38
rs774720586 Chr.5: 176096819-176096819 on Build GRCh38
rs571671940 Chr.5: 176096774-176096774 on Build GRCh38
rs765645368 Chr.5: 176096903-176096903 on Build GRCh38
rs775859254 Chr.5: 176096827-176096827 on Build GRCh38
rs772017788 Chr.5: 176096880-176096880 on Build GRCh38
rs551138043 Chr.5: 176096796-176096796 on Build GRCh38
rs528799445 Chr.5: 176096753-176096753 on Build GRCh38
rs771239903 Chr.5: 176096812-176096812 on Build GRCh38
rs377013685 Chr.5: 176096831-176096831 on Build GRCh38
rs201890428 Chr.5: 176096731-176096731 on Build GRCh38
rs759478059 Chr.5: 176096824-176096824 on Build GRCh38
rs368575316 Chr.5: 176096882-176096882 on Build GRCh38

View Full Product Details