Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 60149448-60149673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS7
Entrez Gene ID
51635
Gene Name
dehydrogenase/reductase 7
Gene Aliases
CGI-86, SDR34C1, retDSR4, retSDR4
UniGene
Hs.59719
Gene Chromosome Location
Chr.14: 60144119-60169864 on Build GRCh38
Transcripts
NM_001322280.1 NM_001322281.1 NM_001322282.1 NM_016029.3 XM_017021368.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3815016 c.873C>T p.F291F Chr.14: 60149452-60149452 on Build GRCh38
3496641 c.819G>A p.M273I Chr.14: 60149506-60149506 on Build GRCh38
348958 c.860C>G p.S287* Chr.14: 60149465-60149465 on Build GRCh38
389135 c.860C>T p.S287L Chr.14: 60149465-60149465 on Build GRCh38
4409140 c.757-2A>G p.? Chr.14: 60149570-60149570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779212069 Chr.14: 60149573-60149573 on Build GRCh38
rs946394402 Chr.14: 60149531-60149531 on Build GRCh38
rs771887533 Chr.14: 60149575-60149575 on Build GRCh38
rs902436805 Chr.14: 60149605-60149605 on Build GRCh38
rs73312144 Chr.14: 60149606-60149606 on Build GRCh38
rs571043736 Chr.14: 60149627-60149627 on Build GRCh38
rs776316620 Chr.14: 60149514-60149514 on Build GRCh38
rs751946309 Chr.14: 60149520-60149520 on Build GRCh38
rs753922867 Chr.14: 60149560-60149560 on Build GRCh38
rs768199302 Chr.14: 60149513-60149513 on Build GRCh38
rs759889970 Chr.14: 60149521-60149521 on Build GRCh38
rs745663724 Chr.14: 60149574-60149574 on Build GRCh38
rs779922120 Chr.14: 60149590-60149590 on Build GRCh38
rs567645047 Chr.14: 60149645-60149645 on Build GRCh38
rs753248149 Chr.14: 60149529-60149529 on Build GRCh38
rs61743926 Chr.14: 60149536-60149536 on Build GRCh38
rs761425805 Chr.14: 60149517-60149517 on Build GRCh38
rs780059429 Chr.14: 60149593-60149593 on Build GRCh38
rs771915686 Chr.14: 60149478-60149478 on Build GRCh38
rs895016552 Chr.14: 60149650-60149650 on Build GRCh38
rs1031639407 Chr.14: 60149635-60149635 on Build GRCh38
rs533715327 Chr.14: 60149639-60149639 on Build GRCh38
rs760170549 Chr.14: 60149509-60149509 on Build GRCh38
rs757586687 Chr.14: 60149564-60149564 on Build GRCh38
rs745607417 Chr.14: 60149475-60149475 on Build GRCh38
rs778880826 Chr.14: 60149474-60149474 on Build GRCh38
rs764832117 Chr.14: 60149519-60149519 on Build GRCh38
rs1056310735 Chr.14: 60149628-60149628 on Build GRCh38
rs111725318 Chr.14: 60149479-60149479 on Build GRCh38
rs756192130 Chr.14: 60149534-60149534 on Build GRCh38
rs539765495 Chr.14: 60149523-60149523 on Build GRCh38

View Full Product Details