Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130012828-130013087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCORL1
Entrez Gene ID
63035
Gene Name
BCL6 corepressor-like 1
Gene Aliases
BCoR-L1, CXorf10
UniGene
Hs.496748
Gene Chromosome Location
Chr.X: 129980285-130058083 on Build GRCh38
Transcripts
NM_001184772.2 NM_021946.4 XM_005262452.4 XM_005262453.4 XM_005262454.3 XM_005262455.4 XM_005262456.4 XM_006724776.3 XM_006724777.3 XM_006724779.2 XM_017029721.1 XM_017029722.1

SNP Detail

SNP ID Chromosome Location
rs973864978 Chr.X: 130012972-130012972 on Build GRCh38
rs779498199 Chr.X: 130012986-130012987 on Build GRCh38
rs745431187 Chr.X: 130013029-130013029 on Build GRCh38
rs181777537 Chr.X: 130012876-130012876 on Build GRCh38
rs141901231 Chr.X: 130012986-130012986 on Build GRCh38
rs770494128 Chr.X: 130012911-130012911 on Build GRCh38
rs1029755096 Chr.X: 130012858-130012858 on Build GRCh38
rs753094647 Chr.X: 130012943-130012943 on Build GRCh38
rs756903033 Chr.X: 130013002-130013002 on Build GRCh38
rs534869537 Chr.X: 130013051-130013051 on Build GRCh38
rs761962656 Chr.X: 130012937-130012937 on Build GRCh38
rs764558968 Chr.X: 130012931-130012931 on Build GRCh38
rs754939100 Chr.X: 130012979-130012979 on Build GRCh38
rs367550651 Chr.X: 130012919-130012919 on Build GRCh38
rs772534414 Chr.X: 130013063-130013063 on Build GRCh38
rs200530257 Chr.X: 130013013-130013013 on Build GRCh38
rs370194529 Chr.X: 130012969-130012969 on Build GRCh38
rs774594848 Chr.X: 130012935-130012935 on Build GRCh38
rs1020902155 Chr.X: 130013010-130013010 on Build GRCh38
rs775984166 Chr.X: 130012914-130012914 on Build GRCh38
rs139887979 Chr.X: 130013052-130013052 on Build GRCh38
rs752641598 Chr.X: 130012981-130012981 on Build GRCh38
rs373755234 Chr.X: 130012905-130012905 on Build GRCh38
rs758119674 Chr.X: 130012982-130012982 on Build GRCh38
rs1006596638 Chr.X: 130012869-130012869 on Build GRCh38
rs1033549152 Chr.X: 130012881-130012881 on Build GRCh38
rs746653501 Chr.X: 130012989-130012989 on Build GRCh38
rs765213234 Chr.X: 130012980-130012980 on Build GRCh38
rs955007181 Chr.X: 130012867-130012867 on Build GRCh38
rs150631012 Chr.X: 130013042-130013042 on Build GRCh38

View Full Product Details