Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34632483-34632754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHF
Entrez Gene ID
26298
Gene Name
ETS homologous factor
Gene Aliases
ESE3, ESE3B, ESEJ
UniGene
Hs.653859
Gene Chromosome Location
Chr.11: 34621041-34663288 on Build GRCh38
Transcripts
NM_001206615.1 NM_001206616.1 NM_012153.5 XM_005252857.1 XM_005252860.1 XM_005252861.1 XM_005252862.1 XM_011519983.1 XM_011519984.1 XM_017017534.1

SNP Detail

SNP ID Chromosome Location
rs978324943 Chr.11: 34632553-34632553 on Build GRCh38
rs191422792 Chr.11: 34632719-34632719 on Build GRCh38
rs554525006 Chr.11: 34632696-34632696 on Build GRCh38
rs780341580 Chr.11: 34632727-34632737 on Build GRCh38
rs1040485907 Chr.11: 34632689-34632689 on Build GRCh38
rs775469102 Chr.11: 34632590-34632591 on Build GRCh38
rs769674224 Chr.11: 34632587-34632590 on Build GRCh38
rs534930675 Chr.11: 34632679-34632679 on Build GRCh38
rs1021397030 Chr.11: 34632508-34632508 on Build GRCh38
rs984511224 Chr.11: 34632727-34632727 on Build GRCh38
rs144745038 Chr.11: 34632610-34632611 on Build GRCh38
rs574094373 Chr.11: 34632697-34632697 on Build GRCh38
rs772234738 Chr.11: 34632601-34632601 on Build GRCh38
rs748143003 Chr.11: 34632707-34632707 on Build GRCh38
rs936800988 Chr.11: 34632606-34632606 on Build GRCh38
rs960283039 Chr.11: 34632633-34632633 on Build GRCh38
rs374830628 Chr.11: 34632603-34632603 on Build GRCh38
rs771103138 Chr.11: 34632726-34632726 on Build GRCh38
rs968857812 Chr.11: 34632531-34632531 on Build GRCh38
rs915928621 Chr.11: 34632686-34632686 on Build GRCh38
rs572121719 Chr.11: 34632618-34632618 on Build GRCh38
rs925456460 Chr.11: 34632602-34632602 on Build GRCh38
rs910260221 Chr.11: 34632611-34632611 on Build GRCh38
rs754793836 Chr.11: 34632539-34632539 on Build GRCh38
rs1032252565 Chr.11: 34632556-34632556 on Build GRCh38
rs763716526 Chr.11: 34632669-34632681 on Build GRCh38
rs993048385 Chr.11: 34632641-34632641 on Build GRCh38

View Full Product Details