Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55678511-55678782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655352 c.132C>T p.A44A Chr.19: 55678759-55678759 on Build GRCh38
1396642 c.448C>A p.L150I Chr.19: 55678742-55678742 on Build GRCh38
1396643 c.115C>A p.L39I Chr.19: 55678742-55678742 on Build GRCh38
1396641 c.301G>A p.A101T Chr.19: 55678595-55678595 on Build GRCh38
6274791 c.267G>T p.S89S Chr.19: 55678561-55678561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766693268 Chr.19: 55678594-55678594 on Build GRCh38
rs752369708 Chr.19: 55678595-55678595 on Build GRCh38
rs35764106 Chr.19: 55678714-55678715 on Build GRCh38
rs763635820 Chr.19: 55678610-55678610 on Build GRCh38
rs542251105 Chr.19: 55678561-55678561 on Build GRCh38
rs773627582 Chr.19: 55678711-55678711 on Build GRCh38
rs1002337792 Chr.19: 55678627-55678627 on Build GRCh38
rs369216651 Chr.19: 55678723-55678723 on Build GRCh38
rs762662519 Chr.19: 55678759-55678759 on Build GRCh38
rs769654129 Chr.19: 55678576-55678576 on Build GRCh38
rs762772140 Chr.19: 55678578-55678579 on Build GRCh38
rs775859328 Chr.19: 55678579-55678579 on Build GRCh38
rs375165854 Chr.19: 55678666-55678666 on Build GRCh38
rs751165306 Chr.19: 55678762-55678762 on Build GRCh38
rs369130195 Chr.19: 55678591-55678591 on Build GRCh38
rs202047340 Chr.19: 55678639-55678639 on Build GRCh38
rs949500865 Chr.19: 55678530-55678530 on Build GRCh38
rs749478444 Chr.19: 55678582-55678582 on Build GRCh38
rs781106190 Chr.19: 55678572-55678572 on Build GRCh38
rs778554427 Chr.19: 55678554-55678554 on Build GRCh38
rs760956596 Chr.19: 55678593-55678593 on Build GRCh38
rs201101921 Chr.19: 55678565-55678565 on Build GRCh38
rs768966602 Chr.19: 55678674-55678674 on Build GRCh38
rs768130917 Chr.19: 55678560-55678560 on Build GRCh38
rs267605689 Chr.19: 55678758-55678758 on Build GRCh38
rs779941442 Chr.19: 55678638-55678638 on Build GRCh38
rs114396589 Chr.19: 55678633-55678633 on Build GRCh38
rs745750069 Chr.19: 55678620-55678620 on Build GRCh38
rs761121951 Chr.19: 55678718-55678718 on Build GRCh38
rs377597073 Chr.19: 55678587-55678587 on Build GRCh38
rs758025416 Chr.19: 55678596-55678596 on Build GRCh38
rs141127387 Chr.19: 55678552-55678552 on Build GRCh38
rs751028974 Chr.19: 55678616-55678616 on Build GRCh38
rs774551624 Chr.19: 55678686-55678686 on Build GRCh38
rs893775327 Chr.19: 55678621-55678621 on Build GRCh38
rs776951435 Chr.19: 55678750-55678750 on Build GRCh38
rs754039807 Chr.19: 55678539-55678539 on Build GRCh38
rs370030834 Chr.19: 55678642-55678642 on Build GRCh38
rs757113261 Chr.19: 55678571-55678571 on Build GRCh38
rs532195098 Chr.19: 55678589-55678589 on Build GRCh38
rs370151707 Chr.19: 55678609-55678609 on Build GRCh38
rs781003404 Chr.19: 55678619-55678619 on Build GRCh38
rs745634936 Chr.19: 55678573-55678573 on Build GRCh38
rs540025424 Chr.19: 55678586-55678586 on Build GRCh38
rs556954533 Chr.19: 55678537-55678537 on Build GRCh38
rs200052849 Chr.19: 55678618-55678618 on Build GRCh38
rs750752386 Chr.19: 55678551-55678551 on Build GRCh38
rs373209112 Chr.19: 55678564-55678564 on Build GRCh38
rs763721618 Chr.19: 55678761-55678761 on Build GRCh38
rs773713771 Chr.19: 55678592-55678592 on Build GRCh38
rs772126895 Chr.19: 55678687-55678687 on Build GRCh38

View Full Product Details